Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dr03074238_m1
See other PTK2AB GE Assays ›
Gene Symbol:
ptk2ab
Gene Name:
protein tyrosine kinase 2ab
Gene Aliases:
fak, fak1a, ptk2, ptk2.1, ptk2a, wu:fa18c04, zgc:66114
Chromosome Location:
Chr.16: 30737278 - 30869776 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID
Dr03074238_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ptk2ab
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_131796.1
NP_571871.1
30 - 31
3071
66
XM_005158162.4
XP_005158219.1
32 - 33
3495
66
XM_005158163.4
XP_005158220.1
31 - 32
3474
66
XM_005158164.4
XP_005158221.1
30 - 31
3456
66
XM_005158165.4
XP_005158222.1
29 - 30
3447
66
XM_009292411.3
XP_009290686.1
31 - 32
3486
66
XM_021466598.1
XP_021322273.1
33 - 34
3611
66
XM_021466599.1
XP_021322274.1
33 - 34
3153
66
XM_021466600.1
XP_021322275.1
33 - 34
3602
66
XM_021466601.1
XP_021322276.1
32 - 33
3601
66
XM_021466602.1
XP_021322277.1
34 - 35
3175
66
XM_021466603.1
XP_021322278.1
32 - 33
3593
66
XM_021466604.1
XP_021322279.1
32 - 33
3583
66
XM_021466605.1
XP_021322280.1
32 - 33
3580
66
XM_021466606.1
XP_021322281.1
31 - 32
3562
66
XM_021466607.1
XP_021322282.1
32 - 33
3505
66
XM_021466608.1
XP_021322283.1
31 - 32
3495
66
XM_021466609.1
XP_021322284.1
34 - 35
3270
66
GenBank mRNA
AF348085.1
-
30 - 31
3071
66
BC163886.1
-
28 - 29
2884
66
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
3' Most
Mammalian Gene Collection
Danio rerio
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
signal complex assembly
epidermal growth factor receptor signaling pathway
phosphorylation
midbrain-hindbrain boundary morphogenesis
cell differentiation
regulation of cell adhesion
peptidyl-tyrosine autophosphorylation
regulation of cell population proliferation
innate immune response
lens fiber cell morphogenesis
protein kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
signaling receptor binding
ATP binding
kinase activity
transferase activity
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0