Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnmt3bb.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025450.1 | NP_001020621.1 | 14 - 15 | 1637 | 82 | |
XM_005162028.4 | XP_005162085.1 | 14 - 15 | 1841 | 82 | ||
XM_009296716.3 | XP_009294991.1 | 13 - 14 | 1695 | 82 | ||
XM_009296717.3 | XP_009294992.1 | 14 - 15 | 1845 | 82 | ||
GenBank mRNA | AB196915.1 | - | 14 - 15 | 1637 | 82 |
Set Membership: |
![]() ![]() ![]() ![]() |