Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fgf17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214808.1 | NP_999973.1 | 4 - 5 | 551 | 73 | |
XM_005172491.4 | XP_005172548.1 | 4 - 5 | 981 | 73 | ||
XM_005172492.4 | XP_005172549.1 | 4 - 5 | 978 | 73 | ||
GenBank mRNA | AY457142.1 | - | 4 - 5 | 551 | 73 | |
BC083269.1 | - | 4 - 5 | 426 | 73 | ||
BC165029.1 | - | 4 - 5 | 396 | 73 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |