Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dr03105657_m1
          See other FGF8A GE Assays ›
          Gene Symbol:
          fgf8a
          Gene Name:
          fibroblast growth factor 8a
          Gene Aliases:
          ace, cb110, etID309886.13, fgf-8, fgf8, id:ibd5031, wu:fb73a06
          Chromosome Location:
          Chr.13: 28495517 - 28501985 on Build GRCz11
          Species:
          Zebrafish
          Species Specific ID (Flybase ID):
          -
          Assay ID Dr03105657_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: fgf8a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_131281.2 NP_571356.2 2 - 3 533 86
          GenBank mRNA AF030560.2 - 2 - 3 457 86
          AF034264.1 - 2 - 3 75 86
          AF051365.1 - 2 - 3 67 86
          BC086718.1 - 2 - 3 533 86
          BC165018.1 - 2 - 3 101 86
          KM655579.1 - 2 - 3 67 86

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          cell fate specification
          somitogenesis
          positive regulation of protein phosphorylation
          heart morphogenesis
          cardiac ventricle morphogenesis
          determination of left/right symmetry
          brain development
          peripheral nervous system development
          endoderm development
          mesoderm development
          positive regulation of cell population proliferation
          fibroblast growth factor receptor signaling pathway
          embryonic pattern specification
          animal organ morphogenesis
          anterior/posterior pattern specification
          dorsal/ventral pattern formation
          cardioblast differentiation
          positive regulation of gene expression
          positive regulation of somitomeric trunk muscle development
          cerebellum development
          midbrain-hindbrain boundary morphogenesis
          rhombomere 5 development
          rhombomere 6 development
          cerebellum formation
          locus ceruleus development
          cell differentiation
          positive regulation of Wnt signaling pathway
          neuron differentiation
          regulation of cell migration
          positive regulation of cell migration
          thyroid gland development
          midbrain development
          hindbrain development
          notochord development
          otic vesicle formation
          midbrain-hindbrain boundary development
          embryonic camera-type eye development
          embryonic heart tube development
          negative regulation of endodermal cell fate specification
          otic placode formation
          regulation of bone remodeling
          determination of dorsal identity
          paraxial mesoderm development
          neuron fate specification
          embryonic neurocranium morphogenesis
          skeletal system morphogenesis
          oligodendrocyte differentiation
          inner ear development
          striated muscle cell differentiation
          cartilage development
          atrial cardiac muscle cell differentiation
          ventricular cardiac muscle cell differentiation
          pharyngeal system development
          retina development in camera-type eye
          embryonic retina morphogenesis in camera-type eye
          cilium assembly
          ectodermal placode formation
          somite development
          taste bud formation
          closure of optic fissure
          Kupffer's vesicle development
          otic vesicle development
          fibroblast growth factor receptor binding
          type 1 fibroblast growth factor receptor binding
          type 2 fibroblast growth factor receptor binding
          growth factor activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0