Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dr03108827_m1
          See other ANK2B GE Assays ›
          Gene Symbol:
          ank2b
          Gene Name:
          ankyrin 2b, neuronal
          Gene Aliases:
          ank2, zgc:101738
          Chromosome Location:
          Chr.7: 57886148 - 58130722 on Build GRCz11
          Species:
          Zebrafish
          Species Specific ID (Flybase ID):
          -
          Assay ID Dr03108827_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ank2b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001346207.1 NP_001333136.1 2 - 3 330 101
          XM_021477473.1 XP_021333148.1 2 - 3 502 101
          XM_021477474.1 XP_021333149.1 2 - 3 502 101
          XM_021477475.1 XP_021333150.1 2 - 3 502 101
          XM_021477477.1 XP_021333152.1 2 - 3 503 101
          XM_021477478.1 XP_021333153.1 2 - 3 378 101
          XM_021477479.1 XP_021333154.1 2 - 3 502 101
          XM_021477480.1 XP_021333155.1 2 - 3 502 101
          XM_021477481.1 XP_021333156.1 2 - 3 502 101
          XM_021477482.1 XP_021333157.1 2 - 3 502 101
          XM_021477483.1 XP_021333158.1 2 - 3 327 101
          XM_021477484.1 XP_021333159.1 2 - 3 502 101
          XM_021477485.1 XP_021333160.1 2 - 3 349 101
          XM_021477486.1 XP_021333161.1 2 - 3 502 101
          XM_021477487.1 XP_021333162.1 2 - 3 502 101
          XM_021477488.1 XP_021333163.1 2 - 3 502 101
          XM_021477489.1 XP_021333164.1 2 - 3 269 101
          XM_021477490.1 XP_021333165.1 2 - 3 502 101
          XM_021477491.1 XP_021333166.1 2 - 3 502 101
          XM_021477492.1 XP_021333167.1 2 - 3 502 101
          XM_021477493.1 XP_021333168.1 2 - 3 502 101
          XM_021477494.1 XP_021333169.1 2 - 3 503 101
          XM_021477495.1 XP_021333170.1 2 - 3 503 101
          XM_021477496.1 XP_021333171.1 2 - 3 502 101
          XM_021477497.1 XP_021333172.1 2 - 3 502 101
          XM_021477498.1 XP_021333173.1 2 - 3 502 101
          XM_021477499.1 XP_021333174.1 2 - 3 502 101
          XM_021477500.1 XP_021333175.1 2 - 3 502 101
          XM_021477501.1 XP_021333176.1 2 - 3 502 101
          XM_021477502.1 XP_021333177.1 2 - 3 503 101
          XM_021477503.1 XP_021333178.1 2 - 3 500 101
          XM_021477504.1 XP_021333179.1 2 - 3 500 101
          XM_021477505.1 XP_021333180.1 2 - 3 500 101
          XM_021477506.1 XP_021333181.1 2 - 3 500 101
          XM_021477507.1 XP_021333182.1 2 - 3 502 101
          XM_021477508.1 XP_021333183.1 2 - 3 500 101
          XM_021477509.1 XP_021333184.1 2 - 3 500 101
          XM_021477510.1 XP_021333185.1 2 - 3 498 101
          XM_021477511.1 XP_021333186.1 2 - 3 512 101
          XM_021477512.1 XP_021333187.1 2 - 3 512 101
          GenBank mRNA BC083261.1 - 2 - 3 323 101

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          signal transduction
          protein localization to T-tubule
          regulation of cardiac muscle contraction
          aspartic-type endopeptidase activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0