Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dr03108830_m1
See other ANK2B GE Assays ›
Gene Symbol:
ank2b
Gene Name:
ankyrin 2b, neuronal
Gene Aliases:
ank2, zgc:101738
Chromosome Location:
Chr.7: 57886148 - 58130722 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID
Dr03108830_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ank2b
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001346207.1
NP_001333136.1
5 - 6
625
85
XM_021477473.1
XP_021333148.1
5 - 6
797
85
XM_021477474.1
XP_021333149.1
5 - 6
797
85
XM_021477475.1
XP_021333150.1
5 - 6
797
85
XM_021477477.1
XP_021333152.1
5 - 6
798
85
XM_021477478.1
XP_021333153.1
5 - 6
673
85
XM_021477479.1
XP_021333154.1
5 - 6
797
85
XM_021477480.1
XP_021333155.1
5 - 6
797
85
XM_021477481.1
XP_021333156.1
5 - 6
797
85
XM_021477482.1
XP_021333157.1
5 - 6
797
85
XM_021477483.1
XP_021333158.1
5 - 6
622
85
XM_021477484.1
XP_021333159.1
5 - 6
797
85
XM_021477485.1
XP_021333160.1
5 - 6
644
85
XM_021477486.1
XP_021333161.1
5 - 6
797
85
XM_021477487.1
XP_021333162.1
5 - 6
797
85
XM_021477488.1
XP_021333163.1
5 - 6
797
85
XM_021477489.1
XP_021333164.1
5 - 6
564
85
XM_021477490.1
XP_021333165.1
5 - 6
797
85
XM_021477491.1
XP_021333166.1
5 - 6
797
85
XM_021477492.1
XP_021333167.1
5 - 6
797
85
XM_021477493.1
XP_021333168.1
5 - 6
797
85
XM_021477494.1
XP_021333169.1
5 - 6
798
85
XM_021477495.1
XP_021333170.1
5 - 6
798
85
XM_021477496.1
XP_021333171.1
5 - 6
797
85
XM_021477497.1
XP_021333172.1
5 - 6
797
85
XM_021477498.1
XP_021333173.1
5 - 6
797
85
XM_021477499.1
XP_021333174.1
5 - 6
797
85
XM_021477500.1
XP_021333175.1
5 - 6
797
85
XM_021477501.1
XP_021333176.1
5 - 6
797
85
XM_021477502.1
XP_021333177.1
5 - 6
798
85
XM_021477503.1
XP_021333178.1
5 - 6
795
85
XM_021477504.1
XP_021333179.1
5 - 6
795
85
XM_021477505.1
XP_021333180.1
5 - 6
795
85
XM_021477506.1
XP_021333181.1
5 - 6
795
85
XM_021477507.1
XP_021333182.1
5 - 6
797
85
XM_021477508.1
XP_021333183.1
5 - 6
795
85
XM_021477509.1
XP_021333184.1
5 - 6
795
85
XM_021477510.1
XP_021333185.1
5 - 6
793
85
XM_021477511.1
XP_021333186.1
5 - 6
807
85
XM_021477512.1
XP_021333187.1
5 - 6
807
85
GenBank mRNA
BC083261.1
-
5 - 6
618
85
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
3' Most
Mammalian Gene Collection
Recommended Assay
Danio rerio
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
signal transduction
protein localization to T-tubule
regulation of cardiac muscle contraction
aspartic-type endopeptidase activity
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0