Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sh3gl3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081691.2 | NP_001075160.1 | 2 - 3 | 285 | 112 | |
GenBank mRNA | BC164395.1 | - | 2 - 3 | 146 | 112 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |