Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dr03112455_m1
See other CASKA GE Assays ›
Gene Symbol:
caska
Gene Name:
calcium/calmodulin-dependent serine protein kinase a
Gene Aliases:
cask, wu:fb70g06
Chromosome Location:
Chr.9: 33365776 - 33609002 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID
Dr03112455_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: caska
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_152941.1
NP_694420.1
2 - 3
685
81
XM_017357621.2
XP_017213110.1
2 - 3
760
81
XM_017357622.2
XP_017213111.1
2 - 3
760
81
XM_017357624.2
XP_017213113.1
2 - 3
761
81
XM_017357625.2
XP_017213114.1
2 - 3
760
81
XM_017357626.2
XP_017213115.1
2 - 3
760
81
XM_017357627.2
XP_017213116.1
2 - 3
759
81
XM_017357629.2
XP_017213118.1
2 - 3
760
81
XM_017357630.2
XP_017213119.1
2 - 3
760
81
XM_017357631.2
XP_017213120.1
2 - 3
760
81
XM_017357632.2
XP_017213121.1
2 - 3
760
81
XM_017357633.2
XP_017213122.1
2 - 3
759
81
XM_017357634.2
XP_017213123.1
2 - 3
759
81
XM_017357635.2
XP_017213124.1
2 - 3
760
81
XM_017357636.2
XP_017213125.1
2 - 3
759
81
XM_017357637.2
XP_017213126.1
2 - 3
760
81
XM_017357638.2
XP_017213127.1
2 - 3
758
81
XM_017357639.2
XP_017213128.1
2 - 3
760
81
XM_017357640.2
XP_017213129.1
2 - 3
759
81
XM_017357641.2
XP_017213130.1
2 - 3
758
81
XM_017357642.2
XP_017213131.1
2 - 3
759
81
XM_017357643.2
XP_017213132.1
2 - 3
758
81
XM_017357644.2
XP_017213133.1
2 - 3
759
81
XM_021478600.1
XP_021334275.1
2 - 3
758
81
GenBank mRNA
AF252546.1
-
2 - 3
685
81
BC162667.1
-
2 - 3
189
81
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
Mammalian Gene Collection
Danio rerio
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
protein kinase activity
ATP binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0