Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dr03112463_m1
          See other CASKA GE Assays ›
          Gene Symbol:
          caska
          Gene Name:
          calcium/calmodulin-dependent serine protein kinase a
          Gene Aliases:
          cask, wu:fb70g06
          Chromosome Location:
          Chr.9: 33365776 - 33609002 on Build GRCz11
          Species:
          Zebrafish
          Species Specific ID (Flybase ID):
          -
          Assay ID Dr03112463_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: caska
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_152941.1 NP_694420.1 6 - 7 1050 96
          XM_017357621.2 XP_017213110.1 6 - 7 1125 96
          XM_017357622.2 XP_017213111.1 6 - 7 1125 96
          XM_017357624.2 XP_017213113.1 6 - 7 1126 96
          XM_017357625.2 XP_017213114.1 6 - 7 1125 96
          XM_017357626.2 XP_017213115.1 6 - 7 1125 96
          XM_017357627.2 XP_017213116.1 6 - 7 1124 96
          XM_017357629.2 XP_017213118.1 6 - 7 1125 96
          XM_017357630.2 XP_017213119.1 6 - 7 1125 96
          XM_017357631.2 XP_017213120.1 6 - 7 1125 96
          XM_017357632.2 XP_017213121.1 6 - 7 1125 96
          XM_017357633.2 XP_017213122.1 6 - 7 1124 96
          XM_017357634.2 XP_017213123.1 6 - 7 1124 96
          XM_017357635.2 XP_017213124.1 6 - 7 1125 96
          XM_017357636.2 XP_017213125.1 6 - 7 1124 96
          XM_017357637.2 XP_017213126.1 6 - 7 1125 96
          XM_017357638.2 XP_017213127.1 6 - 7 1123 96
          XM_017357639.2 XP_017213128.1 6 - 7 1125 96
          XM_017357640.2 XP_017213129.1 6 - 7 1124 96
          XM_017357641.2 XP_017213130.1 6 - 7 1123 96
          XM_017357642.2 XP_017213131.1 6 - 7 1124 96
          XM_017357643.2 XP_017213132.1 6 - 7 1123 96
          XM_017357644.2 XP_017213133.1 6 - 7 1124 96
          XM_021478600.1 XP_021334275.1 6 - 7 1123 96
          GenBank mRNA AF252546.1 - 6 - 7 1050 96
          BC162667.1 - 6 - 7 554 96

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          protein kinase activity
          ATP binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0