Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dr03115946_mH
See other ABLIM1B GE Assays ›
Gene Symbol:
ablim1b
Gene Name:
actin binding LIM protein 1b
Gene Aliases:
si:dkeyp-84e12.1, zgc:112954
Chromosome Location:
Chr.12: 4690 - 44097 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID
Dr03115946_mH
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ablim1b
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_009306613.3
XP_009304888.1
6 - 7
1044
68
XM_009306617.3
XP_009304892.1
6 - 7
1042
68
XM_009306619.3
XP_009304894.1
6 - 7
1041
68
XM_009306622.3
XP_009304897.1
6 - 7
1040
68
XM_009306623.3
XP_009304898.1
6 - 7
1039
68
XM_009306624.3
XP_009304899.1
6 - 7
1041
68
XM_009306627.3
XP_009304902.1
6 - 7
1041
68
XM_017358455.2
XP_017213944.1
6 - 7
1042
68
XM_017358456.2
XP_017213945.1
6 - 7
1040
68
XM_021480065.1
XP_021335740.1
6 - 7
1043
68
XM_021480066.1
XP_021335741.1
6 - 7
1044
68
XM_021480067.1
XP_021335742.1
6 - 7
1043
68
XM_021480068.1
XP_021335743.1
6 - 7
1043
68
XM_021480069.1
XP_021335744.1
6 - 7
1045
68
XM_021480070.1
XP_021335745.1
6 - 7
1044
68
XM_021480071.1
XP_021335746.1
6 - 7
1043
68
XM_021480072.1
XP_021335747.1
6 - 7
1044
68
XM_021480073.1
XP_021335748.1
6 - 7
1043
68
XM_021480074.1
XP_021335749.1
6 - 7
1043
68
XM_021480075.1
XP_021335750.1
6 - 7
1043
68
XM_021480076.1
XP_021335751.1
6 - 7
1043
68
XM_021480078.1
XP_021335753.1
6 - 7
1044
68
XM_021480079.1
XP_021335754.1
6 - 7
1044
68
XM_021480080.1
XP_021335755.1
6 - 7
1045
68
XM_021480081.1
XP_021335756.1
6 - 7
1043
68
XM_021480082.1
XP_021335757.1
6 - 7
1042
68
XM_021480083.1
XP_021335758.1
6 - 7
1043
68
XM_021480084.1
XP_021335759.1
6 - 7
1042
68
XM_021480085.1
XP_021335760.1
6 - 7
1041
68
XM_021480086.1
XP_021335761.1
6 - 7
964
68
XM_021480087.1
XP_021335762.1
6 - 7
835
68
XM_021480088.1
XP_021335763.1
6 - 7
1044
68
XM_021480089.1
XP_021335764.1
6 - 7
1046
68
XM_021480090.1
XP_021335765.1
6 - 7
1044
68
XM_021480091.1
XP_021335766.1
6 - 7
1046
68
XM_021480092.1
XP_021335767.1
6 - 7
1044
68
XM_021480093.1
XP_021335768.1
6 - 7
1043
68
XM_021480094.1
XP_021335769.1
6 - 7
1042
68
XM_021480095.1
XP_021335770.1
6 - 7
1043
68
XM_021480096.1
XP_021335771.1
6 - 7
1042
68
XM_021480097.1
XP_021335772.1
6 - 7
1042
68
XM_021480098.1
XP_021335773.1
6 - 7
1041
68
XM_021480099.1
XP_021335774.1
6 - 7
1051
68
XM_021480100.1
XP_021335775.1
6 - 7
1051
68
XM_021480101.1
XP_021335776.1
6 - 7
1050
68
XM_021480102.1
XP_021335777.1
6 - 7
1050
68
XM_021480104.1
XP_021335779.1
6 - 7
1050
68
GenBank mRNA
BC091932.1
-
6 - 7
873
68
EB891787.1
-
2 - 3
148
68
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
3' Most
Mammalian Gene Collection
Danio rerio
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
lamellipodium assembly
cilium assembly
actin binding
metal ion binding
actin filament binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0