Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: apobb.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030062.1 | NP_001025233.1 | 3 - 4 | 307 | 62 | |
XM_005160629.3 | XP_005160686.1 | 4 - 5 | 410 | 62 |
Set Membership: |
![]() ![]() ![]() ![]() |