Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: apobb.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030062.1 | NP_001025233.1 | 9 - 10 | 1390 | 84 | |
XM_005160629.3 | XP_005160686.1 | 10 - 11 | 1493 | 84 |
Set Membership: |
![]() ![]() ![]() ![]() |