Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dr03130648_m1
          See other PTPRFB GE Assays ›
          Gene Symbol:
          ptprfb
          Gene Name:
          protein tyrosine phosphatase receptor type Fb
          Gene Aliases:
          fj23e12, lar, ptprf, si:dkey-21k10.1, wu:fj23e12
          Chromosome Location:
          Chr.2: 17578415 - 17947246 on Build GRCz11
          Species:
          Zebrafish
          Species Specific ID (Flybase ID):
          -
          Assay ID Dr03130648_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ptprfb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001083576.1 NP_001077045.1 9 - 10 2047 72
          XM_009298457.3 XP_009296732.1 12 - 13 2137 72
          XM_009298458.3 XP_009296733.1 12 - 13 1925 72
          XM_009298459.3 XP_009296734.1 12 - 13 2137 72
          XM_009298460.3 XP_009296735.1 11 - 12 2125 72
          XM_009298461.3 XP_009296736.1 11 - 12 2119 72
          XM_009298462.3 XP_009296737.1 10 - 11 2113 72
          XM_009298463.3 XP_009296738.1 12 - 13 2137 72
          XM_009298464.3 XP_009296739.1 9 - 10 2095 72
          XM_009298466.3 XP_009296741.1 9 - 10 2094 72
          XM_009298467.3 XP_009296742.1 9 - 10 2094 72
          XM_009298468.3 XP_009296743.1 12 - 13 2136 72
          XM_009298469.3 XP_009296744.1 9 - 10 2094 72
          XM_021481410.1 XP_021337085.1 12 - 13 2462 72
          XM_021481419.1 XP_021337094.1 10 - 11 2107 72
          XM_021481437.1 XP_021337112.1 10 - 11 2112 72
          XM_021481438.1 XP_021337113.1 9 - 10 2094 72
          XM_021481440.1 XP_021337115.1 9 - 10 2094 72
          XM_021481441.1 XP_021337116.1 10 - 11 2112 72
          XM_021481443.1 XP_021337118.1 9 - 10 2094 72
          GenBank mRNA BC134804.1 - 9 - 10 2047 72

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          protein dephosphorylation
          cell adhesion
          dephosphorylation
          peptidyl-tyrosine dephosphorylation
          sensory neuron axon guidance
          synaptic membrane adhesion
          phosphoprotein phosphatase activity
          protein tyrosine phosphatase activity
          heparin binding
          hydrolase activity
          phosphatase activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0