Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chmp4bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213457.1 | NP_998622.1 | 3 - 4 | 551 | 79 | |
GenBank mRNA | BC044191.1 | - | 3 - 4 | 551 | 79 | |
BC071537.1 | - | 3 - 4 | 549 | 79 | ||
BC165053.1 | - | 3 - 4 | 506 | 79 |
Set Membership: |
![]() ![]() ![]() ![]() |