Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prdm1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199515.2 | NP_955809.2 | 2 - 3 | 384 | 89 | |
GenBank mRNA | BC080259.1 | - | 2 - 3 | 384 | 89 |
Set Membership: |
![]() ![]() ![]() ![]() |