Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: golga5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213411.2 | NP_998576.1 | 7 - 8 | 1636 | 64 | |
GenBank mRNA | BC055639.1 | - | 7 - 8 | 1638 | 64 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |