Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snrpg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004660.3 | NP_001004660.2 | 2 - 3 | 172 | 84 | |
NM_001303101.1 | NP_001290030.1 | 2 - 3 | 172 | 84 | ||
GenBank mRNA | BC081512.1 | - | 2 - 3 | 147 | 84 | |
BC165520.1 | - | 89 | 84 | |||
EH443016.1 | - | 2 - 3 | 124 | 84 | ||
EH503641.1 | - | 2 - 3 | 117 | 84 | ||
EH549128.1 | - | 2 - 3 | 210 | 84 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |