Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dr03147621_g1
          See other DLG1L GE Assays ›
          Gene Symbol:
          dlg1l
          Gene Name:
          discs, large (Drosophila) homolog 1, like
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 4902770 - 5074670 on Build GRCz11
          Species:
          Zebrafish
          Species Specific ID (Flybase ID):
          -
          Assay ID Dr03147621_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: dlg1l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012388.1 NP_001012388.1 18 - 19 2030 75
          XM_005163667.4 XP_005163724.1 22 - 23 3036 75
          XM_005163668.4 XP_005163725.1 22 - 23 3018 75
          XM_005163669.4 XP_005163726.1 22 - 23 3015 75
          XM_005163671.4 XP_005163728.1 21 - 22 2945 75
          XM_005163673.4 XP_005163730.1 21 - 22 2919 75
          XM_005163675.4 XP_005163732.1 21 - 22 2882 75
          XM_005163676.4 XP_005163733.1 20 - 21 2846 75
          XM_005163677.4 XP_005163734.1 20 - 21 2782 75
          XM_005163678.4 XP_005163735.1 18 - 19 2663 75
          XM_005163682.4 XP_005163739.1 17 - 18 1950 75
          XM_009296932.3 XP_009295207.1 23 - 24 3136 75
          XM_009296934.3 XP_009295209.1 23 - 24 3072 75
          XM_009296940.3 XP_009295215.1 22 - 23 3063 75
          XM_009296951.3 XP_009295226.1 22 - 23 3030 75
          XM_009296964.3 XP_009295239.1 20 - 21 2851 75
          XM_009296971.3 XP_009295246.1 19 - 20 2731 75
          XM_009296979.3 XP_009295254.1 17 - 18 2088 75
          XM_009296993.3 XP_009295268.1 18 - 19 2597 75
          XM_021480105.1 XP_021335780.1 23 - 24 3133 75
          XM_021480123.1 XP_021335798.1 20 - 21 2288 75
          XM_021480126.1 XP_021335801.1 17 - 18 2705 75
          GenBank mRNA AY819035.1 - 18 - 19 2030 75
          BC163455.1 - 18 - 19 2016 75

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          endothelial cell proliferation
          actin filament organization
          chemical synaptic transmission
          embryo development
          cortical actin cytoskeleton organization
          receptor clustering
          establishment or maintenance of epithelial cell apical/basal polarity
          negative regulation of mitotic cell cycle
          receptor localization to synapse
          cell-cell adhesion
          neurotransmitter receptor localization to postsynaptic specialization membrane
          protein kinase binding
          phosphatase binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0