Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dr03147632_m1
          See other DLG1L GE Assays ›
          Gene Symbol:
          dlg1l
          Gene Name:
          discs, large (Drosophila) homolog 1, like
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 4902770 - 5074670 on Build GRCz11
          Species:
          Zebrafish
          Species Specific ID (Flybase ID):
          -
          Assay ID Dr03147632_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: dlg1l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012388.1 NP_001012388.1 9 - 10 1100 69
          XM_005163667.4 XP_005163724.1 12 - 13 2001 69
          XM_005163668.4 XP_005163725.1 12 - 13 1983 69
          XM_005163669.4 XP_005163726.1 12 - 13 1980 69
          XM_005163671.4 XP_005163728.1 11 - 12 1910 69
          XM_005163673.4 XP_005163730.1 11 - 12 1884 69
          XM_005163675.4 XP_005163732.1 11 - 12 1910 69
          XM_005163676.4 XP_005163733.1 10 - 11 1811 69
          XM_005163677.4 XP_005163734.1 10 - 11 1810 69
          XM_005163678.4 XP_005163735.1 8 - 9 1628 69
          XM_005163682.4 XP_005163739.1 7 - 8 915 69
          XM_009296932.3 XP_009295207.1 13 - 14 2101 69
          XM_009296934.3 XP_009295209.1 13 - 14 2100 69
          XM_009296940.3 XP_009295215.1 12 - 13 2028 69
          XM_009296951.3 XP_009295226.1 13 - 14 2100 69
          XM_009296964.3 XP_009295239.1 10 - 11 1816 69
          XM_009296971.3 XP_009295246.1 9 - 10 1696 69
          XM_009296979.3 XP_009295254.1 7 - 8 1053 69
          XM_009296993.3 XP_009295268.1 8 - 9 1625 69
          XM_021480105.1 XP_021335780.1 13 - 14 2098 69
          XM_021480123.1 XP_021335798.1 10 - 11 1253 69
          XM_021480126.1 XP_021335801.1 7 - 8 1670 69
          GenBank mRNA AY819035.1 - 9 - 10 1100 69
          BC163455.1 - 9 - 10 1078 69

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          endothelial cell proliferation
          actin filament organization
          chemical synaptic transmission
          embryo development
          cortical actin cytoskeleton organization
          receptor clustering
          establishment or maintenance of epithelial cell apical/basal polarity
          negative regulation of mitotic cell cycle
          receptor localization to synapse
          cell-cell adhesion
          neurotransmitter receptor localization to postsynaptic specialization membrane
          protein kinase binding
          phosphatase binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0