Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dr03425515_m1
See other ANK3B GE Assays ›
Gene Symbol:
ank3b
Gene Name:
ankyrin 3b
Gene Aliases:
ank3, zgc:171763
Chromosome Location:
Chr.12: 7640825 - 7992115 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID
Dr03425515_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ank3b
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_009306464.3
XP_009304739.1
39 - 40
10649
115
XM_009306467.3
XP_009304742.1
37 - 38
4822
115
XM_009306468.2
XP_009304743.1
36 - 37
4468
115
XM_009306469.3
XP_009304744.1
35 - 36
4738
115
XM_021480224.1
XP_021335899.1
42 - 43
10739
115
XM_021480225.1
XP_021335900.1
42 - 43
10739
115
XM_021480226.1
XP_021335901.1
41 - 42
10727
115
XM_021480227.1
XP_021335902.1
42 - 43
10724
115
XM_021480228.1
XP_021335903.1
41 - 42
10715
115
XM_021480229.1
XP_021335904.1
41 - 42
10712
115
XM_021480230.1
XP_021335905.1
41 - 42
10706
115
XM_021480231.1
XP_021335906.1
42 - 43
10712
115
XM_021480232.1
XP_021335907.1
41 - 42
10694
115
XM_021480233.1
XP_021335908.1
40 - 41
10682
115
XM_021480234.1
XP_021335909.1
41 - 42
10679
115
XM_021480235.1
XP_021335910.1
41 - 42
10679
115
XM_021480236.1
XP_021335911.1
42 - 43
10740
115
XM_021480237.1
XP_021335912.1
43 - 44
10538
115
XM_021480238.1
XP_021335913.1
40 - 41
10667
115
XM_021480239.1
XP_021335914.1
40 - 41
10661
115
XM_021480240.1
XP_021335915.1
42 - 43
10562
115
XM_021480241.1
XP_021335916.1
42 - 43
10739
115
XM_021480242.1
XP_021335917.1
41 - 42
10640
115
XM_021480243.1
XP_021335918.1
42 - 43
10699
115
XM_021480244.1
XP_021335919.1
42 - 43
10350
115
XM_021480245.1
XP_021335920.1
38 - 39
10288
115
XM_021480246.1
XP_021335921.1
38 - 39
10285
115
XM_021480247.1
XP_021335922.1
42 - 43
10740
115
XM_021480248.1
XP_021335923.1
38 - 39
10589
115
XM_021480249.1
XP_021335924.1
37 - 38
10535
115
XM_021480250.1
XP_021335925.1
42 - 43
10739
115
XM_021480251.1
XP_021335926.1
42 - 43
10739
115
XM_021480252.1
XP_021335927.1
42 - 43
10741
115
XM_021480253.1
XP_021335928.1
42 - 43
10741
115
XM_021480254.1
XP_021335929.1
19 - 20
8173
115
XM_021480255.1
XP_021335930.1
18 - 19
8161
115
XM_021480256.1
XP_021335931.1
17 - 18
8116
115
XM_021480257.1
XP_021335932.1
16 - 17
8082
115
XM_021480258.1
XP_021335933.1
41 - 42
4940
115
XM_021480259.1
XP_021335934.1
40 - 41
4913
115
XM_021480260.1
XP_021335935.1
39 - 40
4880
115
GenBank mRNA
BC151969.1
-
1 - 2
282
115
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
Mammalian Gene Collection
Recommended Assay
Danio rerio
Probe spans exons
Gene Ontology Categories:
Function(s)
signal transduction
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0