Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dr03432826_m1
          See other SMAD5 GE Assays ›
          Gene Symbol:
          smad5
          Gene Name:
          SMAD family member 5
          Gene Aliases:
          SBN, fb67b04, mad, madh5, pgy, sb:cb400, smd5, wu:fb67b04
          Chromosome Location:
          Chr.14: 26467200 - 26498207 on Build GRCz11
          Species:
          Zebrafish
          Species Specific ID (Flybase ID):
          -
          Assay ID Dr03432826_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: smad5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001346309.1 NP_001333238.1 4 - 5 1287 67
          NM_131368.3 NP_571443.3 5 - 6 1372 67
          XM_005173164.4 XP_005173221.1 5 - 6 1366 67
          GenBank mRNA AF127920.1 - 3 - 4 1142 67
          AF168775.1 - 3 - 4 777 67
          BC050481.1 - 5 - 6 1362 67
          BC065644.1 - 5 - 6 1254 67
          BC154311.1 - 3 - 4 871 67
          BC155776.1 - 4 - 5 1254 67
          BC165695.1 - 3 - 4 811 67
          DT073830.1 - 3 - 4 424 67

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          lymph vessel development
          heart looping
          embryonic heart tube morphogenesis
          regulation of transcription, DNA-templated
          transforming growth factor beta receptor signaling pathway
          multicellular organism development
          determination of left/right symmetry
          anatomical structure morphogenesis
          embryonic pattern specification
          dorsal/ventral pattern formation
          cell differentiation
          BMP signaling pathway
          post-anal tail morphogenesis
          otic placode formation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          determination of dorsal/ventral asymmetry
          determination of ventral identity
          blood vessel morphogenesis
          posterior lateral line neuromast development
          posterior lateral line neuromast deposition
          roof of mouth development
          dorsal convergence
          pharyngeal system development
          SMAD protein signal transduction
          regulation of BMP signaling pathway involved in heart jogging
          transcription cis-regulatory region binding
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          DNA-binding transcription activator activity, RNA polymerase II-specific
          DNA binding
          DNA-binding transcription factor activity
          protein binding
          transcription factor binding
          ubiquitin protein ligase binding
          metal ion binding
          I-SMAD binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc