Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dr03435728_m1
          See other VEGFAA GE Assays ›
          Gene Symbol:
          vegfaa
          Gene Name:
          vascular endothelial growth factor Aa
          Gene Aliases:
          vegf, vegfa, wu:fj82c06
          Chromosome Location:
          Chr.16: 4287539 - 4317955 on Build GRCz11
          Species:
          Zebrafish
          Species Specific ID (Flybase ID):
          -
          Assay ID Dr03435728_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: vegfaa
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001110349.2 NP_001103819.2 4 - 5 384 66
          NM_131408.3 NP_571483.1 4 - 5 384 66
          XM_009292018.3 XP_009290293.1 4 - 5 1045 66
          GenBank mRNA AF016244.1 - 4 - 5 969 66
          AF059661.1 - 4 - 5 384 66
          BC162258.1 - 4 - 5 429 66
          BC162588.1 - 4 - 5 429 66

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          blood vessel development
          branching involved in blood vessel morphogenesis
          vasculogenesis
          response to hypoxia
          positive regulation of protein phosphorylation
          positive regulation of endothelial cell proliferation
          vasculature development
          sprouting angiogenesis
          blood vessel endothelial cell proliferation involved in sprouting angiogenesis
          endocardial progenitor cell migration to the midline involved in heart field formation
          cardioblast migration to the midline involved in heart rudiment formation
          activation of phospholipase C activity
          multicellular organism development
          regulation of heart contraction
          motor neuron axon guidance
          response to glucose
          hemopoiesis
          cell differentiation
          thyroid gland development
          positive regulation of protein-containing complex assembly
          regulation of angioblast cell migration involved in selective angioblast sprouting
          angioblast cell migration from lateral mesoderm to midline
          vascular endothelial growth factor signaling pathway
          protein kinase B signaling
          positive regulation of angiogenesis
          vascular endothelial growth factor receptor signaling pathway
          pronephros development
          artery morphogenesis
          positive regulation of peptidyl-tyrosine phosphorylation
          induction of positive chemotaxis
          positive regulation of cell division
          positive regulation of focal adhesion assembly
          positive regulation of mast cell chemotaxis
          regulation of vasculature development
          vascular endothelial growth factor receptor binding
          protein binding
          growth factor activity
          heparin binding
          chemoattractant activity
          vascular endothelial growth factor receptor 1 binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-tbs5g:80/100.66.134.240:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0