Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dr03437961_mH
          See other PICALMA GE Assays ›
          Gene Symbol:
          picalma
          Gene Name:
          phosphatidylinositol binding clathrin assembly protein a
          Gene Aliases:
          picalm, zgc:55615
          Chromosome Location:
          Chr.10: 149546 - 238526 on Build GRCz11
          Species:
          Zebrafish
          Species Specific ID (Flybase ID):
          -
          Assay ID Dr03437961_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: picalma
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001365260.1 NP_001352189.1 20 - 21 1990 79
          NM_001365261.1 NP_001352190.1 18 - 19 1816 79
          NM_200927.2 NP_957221.2 17 - 18 1771 79
          XM_009305372.3 XP_009303647.1 22 - 23 2244 79
          XM_009305375.3 XP_009303650.1 22 - 23 2232 79
          XM_009305376.3 XP_009303651.1 22 - 23 2211 79
          XM_009305379.3 XP_009303654.1 22 - 23 2244 79
          XM_009305387.3 XP_009303662.1 18 - 19 1902 79
          XM_009305388.3 XP_009303663.1 18 - 19 1881 79
          XM_009305389.3 XP_009303664.1 18 - 19 1881 79
          XM_009305392.3 XP_009303667.1 21 - 22 2220 79
          XM_021479123.1 XP_021334798.1 23 - 24 2347 79
          XM_021479124.1 XP_021334799.1 23 - 24 2343 79
          XM_021479125.1 XP_021334800.1 22 - 23 2334 79
          XM_021479126.1 XP_021334801.1 23 - 24 2335 79
          XM_021479127.1 XP_021334802.1 23 - 24 2313 79
          XM_021479128.1 XP_021334803.1 23 - 24 2268 79
          XM_021479129.1 XP_021334804.1 22 - 23 2262 79
          XM_021479130.1 XP_021334805.1 21 - 22 2031 79
          XM_021479131.1 XP_021334806.1 19 - 20 1956 79
          XM_021479132.1 XP_021334807.1 20 - 21 1950 79
          XM_021479133.1 XP_021334808.1 19 - 20 1938 79
          XM_021479134.1 XP_021334809.1 20 - 21 1938 79
          XM_021479135.1 XP_021334810.1 19 - 20 1926 79
          XM_021479136.1 XP_021334811.1 20 - 21 1917 79
          XM_021479137.1 XP_021334812.1 19 - 20 1905 79
          XM_021479138.1 XP_021334813.1 22 - 23 2322 79
          GenBank mRNA BC045405.1 - 17 - 18 1731 79

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          endocytosis
          vesicle budding from membrane
          synaptic vesicle budding from presynaptic endocytic zone membrane
          clathrin coat assembly
          synaptic vesicle endocytosis
          clathrin-dependent endocytosis
          SNARE binding
          phospholipid binding
          1-phosphatidylinositol binding
          phosphatidylinositol-4,5-bisphosphate binding
          clathrin binding
          clathrin heavy chain binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline