Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dr03437969_mH
See other PICALMA GE Assays ›
Gene Symbol:
picalma
Gene Name:
phosphatidylinositol binding clathrin assembly protein a
Gene Aliases:
picalm, zgc:55615
Chromosome Location:
Chr.10: 149546 - 238526 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID
Dr03437969_mH
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: picalma
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001365260.1
NP_001352189.1
11 - 12
1121
90
NM_001365261.1
NP_001352190.1
9 - 10
1016
90
NM_200927.2
NP_957221.2
9 - 10
1016
90
XM_009305372.3
XP_009303647.1
11 - 12
1138
90
XM_009305375.3
XP_009303650.1
11 - 12
1138
90
XM_009305376.3
XP_009303651.1
11 - 12
1138
90
XM_009305379.3
XP_009303654.1
11 - 12
1138
90
XM_009305387.3
XP_009303662.1
10 - 11
1126
90
XM_009305388.3
XP_009303663.1
10 - 11
1126
90
XM_009305389.3
XP_009303664.1
10 - 11
1126
90
XM_009305390.3
XP_009303665.1
11 - 12
1219
90
XM_009305392.3
XP_009303667.1
10 - 11
1126
90
XM_021479123.1
XP_021334798.1
11 - 12
1217
90
XM_021479124.1
XP_021334799.1
11 - 12
1219
90
XM_021479125.1
XP_021334800.1
10 - 11
1204
90
XM_021479126.1
XP_021334801.1
11 - 12
1217
90
XM_021479127.1
XP_021334802.1
11 - 12
1216
90
XM_021479128.1
XP_021334803.1
11 - 12
1138
90
XM_021479129.1
XP_021334804.1
11 - 12
1216
90
XM_021479130.1
XP_021334805.1
11 - 12
1138
90
XM_021479131.1
XP_021334806.1
10 - 11
1126
90
XM_021479132.1
XP_021334807.1
11 - 12
1138
90
XM_021479133.1
XP_021334808.1
10 - 11
1126
90
XM_021479134.1
XP_021334809.1
11 - 12
1138
90
XM_021479135.1
XP_021334810.1
10 - 11
1126
90
XM_021479136.1
XP_021334811.1
11 - 12
1138
90
XM_021479137.1
XP_021334812.1
10 - 11
1126
90
XM_021479138.1
XP_021334813.1
10 - 11
1204
90
GenBank mRNA
BC045405.1
-
9 - 10
976
90
BC154792.1
-
10 - 11
1045
90
Back To Top
More Information
Set Membership:
Amplicon length between 86 and 100
Mammalian Gene Collection
Danio rerio
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
vesicle budding from membrane
synaptic vesicle budding from presynaptic endocytic zone membrane
clathrin coat assembly
synaptic vesicle endocytosis
clathrin-dependent endocytosis
SNARE binding
phospholipid binding
1-phosphatidylinositol binding
phosphatidylinositol-4,5-bisphosphate binding
clathrin binding
clathrin heavy chain binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0