Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014738414.2 | XP_014593900.1 | 372 | 94 | ||
XM_014738415.2 | XP_014593901.1 | 266 | 94 | |||
XM_014738416.2 | XP_014593902.1 | 195 | 94 | |||
XM_023637273.1 | XP_023493041.1 | 319 | 94 | |||
XM_023637274.1 | XP_023493042.1 | 471 | 94 | |||
XM_023637278.1 | XP_023493046.1 | 584 | 94 | |||
XM_023637279.1 | XP_023493047.1 | 584 | 94 | |||
XM_023637280.1 | XP_023493048.1 | 584 | 94 | |||
XM_023637281.1 | XP_023493049.1 | 584 | 94 |
Set Membership: |
![]() ![]() ![]() |