Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100062210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005602712.3 | XP_005602769.2 | 2436 | 105 | ||
XM_023647955.1 | XP_023503723.1 | 2433 | 105 | |||
XM_023647957.1 | XP_023503725.1 | 2436 | 105 |
Set Membership: |
![]() ![]() |