Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Gg03347182_m1
          See other ATVR1 GE Assays ›
          Gene Symbol:
          ATVR1
          Gene Name:
          serine/threonine-protein kinase receptor
          Gene Aliases:
          ACVR1, ALK2
          Chromosome Location:
          Chr.7: 36259181 - 36304118 on Build GRCg6a
          Species:
          Chicken
          Species Specific ID (Flybase ID):
          -
          Assay ID Gg03347182_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ATVR1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_204560.1 NP_989891.1 6 - 7 922 68
          XM_015290126.2 XP_015145612.1 6 - 7 1043 68
          XM_015290127.2 XP_015145613.1 6 - 7 874 68
          XM_015290128.2 XP_015145614.1 6 - 7 824 68
          XM_015290130.2 XP_015145616.1 6 - 7 882 68
          XM_015290131.2 XP_015145617.1 6 - 7 877 68
          XM_015290132.2 XP_015145618.1 6 - 7 906 68
          XM_015290133.2 XP_015145619.1 6 - 7 817 68
          XM_015290134.2 XP_015145620.1 6 - 7 871 68
          XM_025152083.1 XP_025007851.1 6 - 7 945 68
          XM_025152084.1 XP_025007852.1 6 - 7 821 68
          GenBank mRNA AJ318064.2 - 6 - 7 922 68
          U38622.1 - 6 - 7 884 68

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Gallus gallus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          angiogenesis
          branching involved in blood vessel morphogenesis
          gastrulation with mouth forming second
          mesoderm formation
          neural crest cell migration
          acute inflammatory response
          embryonic heart tube morphogenesis
          atrioventricular valve morphogenesis
          mitral valve morphogenesis
          endocardial cushion morphogenesis
          endocardial cushion fusion
          atrial septum primum morphogenesis
          protein phosphorylation
          transforming growth factor beta receptor signaling pathway
          germ cell development
          determination of left/right symmetry
          gastrulation
          mesoderm development
          heart development
          dorsal/ventral pattern formation
          negative regulation of signal transduction
          positive regulation of alkaline phosphatase activity
          positive regulation of epithelial to mesenchymal transition
          positive regulation of pathway-restricted SMAD protein phosphorylation
          cell migration
          peptidyl-threonine phosphorylation
          regulation of ossification
          positive regulation of cell migration
          positive regulation of bone mineralization
          BMP signaling pathway
          positive regulation of BMP signaling pathway
          activin receptor signaling pathway
          negative regulation of activin receptor signaling pathway
          intracellular signal transduction
          endothelial cell activation
          positive regulation of osteoblast differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          mesenchymal cell differentiation
          smooth muscle cell differentiation
          pharyngeal system development
          cardiac epithelial to mesenchymal transition
          pathway-restricted SMAD protein phosphorylation
          ventricular septum morphogenesis
          cardiac muscle cell fate commitment
          BMP signaling pathway involved in heart development
          endocardial cushion cell fate commitment
          cellular response to growth factor stimulus
          cellular response to BMP stimulus
          positive regulation of epithelial to mesenchymal transition involved in endocardial cushion formation
          positive regulation of determination of dorsal identity
          negative regulation of extrinsic apoptotic signaling pathway
          protein kinase activity
          protein serine/threonine kinase activity
          transforming growth factor beta receptor activity, type I
          protein binding
          ATP binding
          activin receptor activity, type I
          growth factor binding
          protein homodimerization activity
          SMAD binding
          metal ion binding
          activin binding
          transforming growth factor beta binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0