Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XKR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034824.1 | NP_001029996.1 | 2 - 3 | 524 | 151 | |
XM_015282822.2 | XP_015138308.2 | 4 - 5 | 2725 | 151 | ||
XM_015282823.2 | XP_015138309.2 | 3 - 4 | 915 | 151 | ||
XM_015282824.2 | XP_015138310.2 | 2 - 3 | 1886 | 151 | ||
XM_025147315.1 | XP_025003083.1 | 3 - 4 | 374 | 151 | ||
GenBank mRNA | AY699714.1 | - | 2 - 3 | 524 | 151 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |