Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Gg03365817_m1
          See other HNRNPH1 GE Assays ›
          Gene Symbol:
          HNRNPH1
          Gene Name:
          heterogeneous nuclear ribonucleoprotein H1
          Gene Aliases:
          HNRNPH2, HNRPH1
          Chromosome Location:
          Chr.13: 14281474 - 14298029 on Build GRCg6a
          Species:
          Chicken
          Species Specific ID (Flybase ID):
          -
          Assay ID Gg03365817_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HNRNPH1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_204496.1 NP_989827.1 12 - 13 1560 62
          XM_015293722.2 XP_015149208.2 11 - 12 1507 62
          XM_015293723.2 XP_015149209.2 13 - 14 1576 62
          XM_015293725.2 XP_015149211.2 13 - 14 1679 62
          XM_015293726.2 XP_015149212.2 12 - 13 1516 62
          XM_015293728.2 XP_015149214.2 12 - 13 1421 62
          XM_015293730.2 XP_015149216.2 11 - 12 1488 62
          XM_025154783.1 XP_025010551.1 12 - 13 1587 62
          XM_025154784.1 XP_025010552.1 13 - 14 1556 62
          XM_025154785.1 XP_025010553.1 12 - 13 1581 62
          XM_025154786.1 XP_025010554.1 12 - 13 1376 62
          XM_025154787.1 XP_025010555.1 12 - 13 1569 62
          XM_025154788.1 XP_025010556.1 12 - 13 1560 62
          XM_025154789.1 XP_025010557.1 12 - 13 1569 62
          XM_025154790.1 XP_025010558.1 13 - 14 1465 62
          XM_025154791.1 XP_025010559.1 11 - 12 1528 62
          XM_025154792.1 XP_025010560.1 12 - 13 1495 62
          XM_025154793.1 XP_025010561.1 11 - 12 1518 62
          XM_025154794.1 XP_025010562.1 11 - 12 1480 62
          XM_025154795.1 XP_025010563.1 11 - 12 1467 62
          XM_025154796.1 XP_025010564.1 12 - 13 1385 62
          XM_025154797.1 XP_025010565.1 13 - 14 1652 62
          GenBank mRNA AJ719489.1 - 11 - 12 1476 62
          AJ851662.1 - 12 - 13 1461 62
          AY278548.1 - 12 - 13 1560 62

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix