Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00167241_m1
          See other HTR2A GE Assays ›
          Gene Symbol:
          HTR2A
          Gene Name:
          5-hydroxytryptamine receptor 2A
          Gene Aliases:
          5-HT2A, HTR2
          Chromosome Location:
          Chr.13: 46831542 - 46897076 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00167241_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HTR2A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000621.4 NP_000612.1 2 - 3 1145 77
          GenBank mRNA A23337.1 - 1 - 2 621 77
          AF498982.1 - 1 - 2 414 77
          AK314132.1 - 2 - 3 902 77
          BC069356.1 - 1 - 2 493 7262350 77
          BC069576.1 - 1 - 2 493 7262362 77
          BC074848.2 - 1 - 2 441 30915554 77
          BC074849.2 - 1 - 2 441 30915332 77
          BC096839.1 - 1 - 2 415 6519217 77
          M86841.1 - 1 - 2 390 77
          S71229.1 - 1 - 2 464 77
          X57830.1 - 1 - 2 559 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1300008858.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC13000648.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Homo sapiens Human Neurotransmitters HUMAN INTRACELLULAR CALCIUM SIGNALING HUMAN SIGNALING IN GAP JUNCTIONS Inventoried Human_Neurotransmitter Human_Intracellular_Calcium_Signaling Human_Signaling_in_GAP_Junctions Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HTR2A

          Gene Ontology Categories:

          Function(s) Process(es)

          temperature homeostasis
          cellular calcium ion homeostasis
          activation of phospholipase C activity
          phospholipase C-activating serotonin receptor signaling pathway
          serotonin receptor signaling pathway
          chemical synaptic transmission
          aging
          memory
          cell death
          positive regulation of cell population proliferation
          response to xenobiotic stimulus
          positive regulation of phosphatidylinositol biosynthetic process
          regulation of dopamine secretion
          phosphatidylinositol 3-kinase signaling
          artery smooth muscle contraction
          urinary bladder smooth muscle contraction
          sleep
          negative regulation of potassium ion transport
          positive regulation of MAP kinase activity
          protein localization to cytoskeleton
          positive regulation of fat cell differentiation
          positive regulation of glycolytic process
          positive regulation of vasoconstriction
          viral entry into host cell
          regulation of hormone secretion
          behavioral response to cocaine
          positive regulation of peptidyl-tyrosine phosphorylation
          regulation of behavior
          detection of temperature stimulus involved in sensory perception of pain
          detection of mechanical stimulus involved in sensory perception of pain
          release of sequestered calcium ion into cytosol
          negative regulation of synaptic transmission, glutamatergic
          positive regulation of ERK1 and ERK2 cascade
          virus receptor activity
          G-protein alpha-subunit binding
          G protein-coupled serotonin receptor activity
          protein-containing complex binding
          serotonin binding
          1-(4-iodo-2,5-dimethoxyphenyl)propan-2-amine binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR13007401.hg.1 ENST00000378688.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13007401.hg.1 ENST00000542664.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13007401.hg.1 ENST00000543956.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13007401.hg.1 NM_000621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13007401.hg.1 NM_001165947.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13007401.hg.1 TC13000648.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000543956.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000543956.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000543956.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000543956.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000543956.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000543956.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000543956.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000612998.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000612998.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 ENST00000612998.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 NM_001165947.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 NM_001165947.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 NM_001165947.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 NM_001165947.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 NM_001165947.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 NM_001165947.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008858.hg.1 NM_001165947.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300159829.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1300159829.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1300159829.hg.1 ENST00000612998.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300159829.hg.1 NM_000621.4 Refseq
          Affymetrix Human Clariom D Assay PSR1300159829.hg.1 TC1300008858.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300159829.hg.1 NM_000621.4 Refseq
          Affymetrix Human Clariom D Assay PSR1300159829.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1300159829.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1300159829.hg.1 ENST00000612998.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000543956.4 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000543956.4 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000543956.4 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000543956.4 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000543956.4 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000543956.4 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000543956.4 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000612998.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000612998.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 ENST00000612998.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 NM_000621.4 Refseq
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 NM_001165947.2 Refseq
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 NM_001165947.2 Refseq
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 NM_001165947.2 Refseq
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 NM_001165947.2 Refseq
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 NM_001165947.2 Refseq
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 NM_001165947.2 Refseq
          Affymetrix Human Clariom D Assay TC1300008858.hg.1 NM_001165947.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300159828.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1300159828.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1300159828.hg.1 ENST00000612998.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300159828.hg.1 NM_000621.4 Refseq
          Affymetrix Human Clariom D Assay PSR1300159828.hg.1 TC1300008858.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300159828.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1300159828.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1300159828.hg.1 ENST00000612998.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300159828.hg.1 NM_000621.4 Refseq
          Affymetrix Human Clariom D Assay PSR1300159824.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1300159824.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1300159824.hg.1 ENST00000543956.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1300159824.hg.1 NM_000621.4 Refseq
          Affymetrix Human Clariom D Assay PSR1300159824.hg.1 NM_001165947.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300159824.hg.1 TC1300008858.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300159824.hg.1 ENST00000378688.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1300159824.hg.1 ENST00000542664.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1300159824.hg.1 ENST00000543956.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1300159824.hg.1 NM_000621.4 Refseq
          Affymetrix Human Clariom D Assay PSR1300159824.hg.1 NM_001165947.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0