Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00171132_m1
          See other GDF15 GE Assays ›
          Gene Symbol:
          GDF15
          Gene Name:
          growth differentiation factor 15
          Gene Aliases:
          GDF-15, MIC-1, MIC1, NAG-1, PDF, PLAB, PTGFB
          Chromosome Location:
          Chr.19: 18386158 - 18389176 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00171132_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GDF15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004864.2 NP_004855.2 1 - 2 313 78
          GenBank mRNA AB000584.1 - 1 - 2 313 78
          AF003934.1 - 1 - 2 315 78
          AF019770.1 - 1 - 2 298 78
          AK225431.1 - 1 - 2 313 78
          AK291530.1 - 1 - 2 313 78
          BC000529.2 - 1 - 2 292 2961614 78
          BC008962.2 - 1 - 2 298 3009858 78
          BT019465.1 - 1 - 2 281 78
          U51731.1 - 1 - 2 293 78
          U88323.1 - 1 - 2 306 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900007384.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19000356.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens Human TGFB Panel 1700 Inventoried Human_TGFB Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GDF15

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          transforming growth factor beta receptor signaling pathway
          cell-cell signaling
          positive regulation of pathway-restricted SMAD protein phosphorylation
          regulation of apoptotic process
          regulation of MAPK cascade
          cell development
          SMAD protein signal transduction
          positive regulation of myoblast fusion
          cytokine activity
          transforming growth factor beta receptor binding
          growth factor activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR19004731.hg.1 ENST00000252809.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19004731.hg.1 NM_004864.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19004731.hg.1 TC19000356.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19004732.hg.1 ENST00000252809.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19004732.hg.1 NM_004864.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19004732.hg.1 TC19000356.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1900007384.hg.1 ENST00000252809.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007384.hg.1 ENST00000252809.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007384.hg.1 ENST00000252809.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007384.hg.1 ENST00000252809.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007384.hg.1 ENST00000252809.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007384.hg.1 ENST00000594925.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007384.hg.1 ENST00000594925.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007384.hg.1 ENST00000594925.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007384.hg.1 ENST00000595973.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007384.hg.1 ENST00000595973.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007384.hg.1 ENST00000595973.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007384.hg.1 ENST00000595973.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007384.hg.1 ENST00000595973.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007384.hg.1 ENST00000595973.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007384.hg.1 ENST00000597765.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007384.hg.1 ENST00000597765.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007384.hg.1 ENST00000597765.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007384.hg.1 ENST00000604609.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007384.hg.1 ENST00000604609.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007384.hg.1 ENST00000604609.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007384.hg.1 NM_004864.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007384.hg.1 NM_004864.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007384.hg.1 NM_004864.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007384.hg.1 NM_004864.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007384.hg.1 NM_004864.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900154187.hg.1 ENST00000252809.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1900154187.hg.1 ENST00000594925.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900154187.hg.1 ENST00000595973.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900154187.hg.1 NM_004864.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900154187.hg.1 TC1900007384.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900154187.hg.1 ENST00000252809.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1900154187.hg.1 ENST00000594925.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900154187.hg.1 ENST00000595973.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900154187.hg.1 NM_004864.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900154188.hg.1 ENST00000252809.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1900154188.hg.1 ENST00000594925.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900154188.hg.1 NM_004864.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900154188.hg.1 TC1900007384.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900154188.hg.1 ENST00000252809.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1900154188.hg.1 ENST00000594925.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900154188.hg.1 NM_004864.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900154189.hg.1 ENST00000252809.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1900154189.hg.1 NM_004864.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900154189.hg.1 TC1900007384.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900154189.hg.1 ENST00000252809.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1900154189.hg.1 NM_004864.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007384.hg.1 ENST00000252809.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900007384.hg.1 ENST00000252809.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900007384.hg.1 ENST00000252809.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900007384.hg.1 ENST00000252809.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900007384.hg.1 ENST00000252809.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900007384.hg.1 ENST00000594925.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007384.hg.1 ENST00000594925.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007384.hg.1 ENST00000594925.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007384.hg.1 ENST00000595973.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900007384.hg.1 ENST00000595973.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900007384.hg.1 ENST00000595973.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900007384.hg.1 ENST00000595973.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900007384.hg.1 ENST00000595973.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900007384.hg.1 ENST00000595973.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900007384.hg.1 ENST00000597765.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007384.hg.1 ENST00000597765.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007384.hg.1 ENST00000597765.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007384.hg.1 ENST00000604609.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900007384.hg.1 ENST00000604609.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900007384.hg.1 ENST00000604609.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900007384.hg.1 NM_004864.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007384.hg.1 NM_004864.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007384.hg.1 NM_004864.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007384.hg.1 NM_004864.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007384.hg.1 NM_004864.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900154185.hg.1 ENST00000252809.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1900154185.hg.1 ENST00000595973.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900154185.hg.1 NM_004864.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900154185.hg.1 TC1900007384.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900154185.hg.1 ENST00000252809.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1900154185.hg.1 ENST00000595973.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900154185.hg.1 NM_004864.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 182404 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0