Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012432.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012432.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012432.hg.1 |
TC03000693.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012431.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012431.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012431.hg.1 |
TC03000693.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012429.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012429.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012429.hg.1 |
TC03000693.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012428.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012428.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012428.hg.1 |
TC03000693.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0300162514.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162514.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162514.hg.1 |
TC0300008787.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300162514.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162514.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162516.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162516.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162516.hg.1 |
TC0300008787.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300162516.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162516.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162518.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162518.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162518.hg.1 |
TC0300008787.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300162518.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162518.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162517.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162517.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162517.hg.1 |
TC0300008787.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300162517.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162517.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162513.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162513.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162513.hg.1 |
TC0300008787.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300162513.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162513.hg.1 |
NM_000539.3 |
Refseq |