Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00182411_m1
          See other TNFSF4 GE Assays ›
          Gene Symbol:
          TNFSF4
          Gene Name:
          tumor necrosis factor superfamily member 4
          Gene Aliases:
          CD134L, CD252, GP34, OX-40L, OX4OL, TNLG2B, TXGP1
          Chromosome Location:
          Chr.1: 173183729 - 173462208 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00182411_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TNFSF4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003326.4 NP_003317.1 1 - 2 289 72
          XM_011509964.2 XP_011508266.2 2 - 3 1092 72
          XM_017002229.1 XP_016857718.1 3 - 4 6412 72
          XM_017002230.1 XP_016857719.1 2 - 3 776 72
          GenBank mRNA D90224.1 - 1 - 2 188 72
          X79929.1 - 1 - 2 289 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100016431.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01003525.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Immune Tolerance Network Homo sapiens Human TNF Superfamily Pathway Inventoried Human_TNF_Superfamily_Pathway Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TNFSF4

          Gene Ontology Categories:

          Function(s) Process(es)

          defense response to nematode
          acute inflammatory response
          positive regulation of T cell cytokine production
          regulation of adaptive immune response
          positive regulation of type 2 immune response
          positive regulation of immunoglobulin mediated immune response
          immune response
          signal transduction
          cholesterol metabolic process
          positive regulation of cell proliferation
          response to virus
          negative regulation of interferon-gamma production
          negative regulation of interleukin-17 production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-10 production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-13 production
          positive regulation of interleukin-2 production
          positive regulation of interleukin-4 production
          positive regulation of interleukin-6 production
          tumor necrosis factor-mediated signaling pathway
          memory T cell activation
          T-helper 2 cell activation
          response to nitrogen dioxide
          cellular response to nitrogen dioxide
          CD4-positive, alpha-beta T cell costimulation
          T cell proliferation
          positive regulation of T cell proliferation
          positive regulation of activated T cell proliferation
          positive regulation of CD4-positive, alpha-beta T cell differentiation
          positive regulation of memory T cell differentiation
          negative regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of regulatory T cell differentiation
          negative regulation of T-helper 1 cell differentiation
          positive regulation of T-helper 2 cell differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of alpha-beta T cell proliferation
          negative regulation of cytokine secretion
          regulation of inflammatory response
          positive regulation of inflammatory response
          positive regulation of B cell activation
          positive regulation of immunoglobulin secretion
          cellular response to lipopolysaccharide
          cellular response to prostaglandin E stimulus
          chemokine (C-C motif) ligand 11 production
          positive regulation of CD4-positive, alpha-beta T cell costimulation
          positive regulation of memory T cell activation
          positive regulation of T-helper 2 cell activation
          positive regulation of interleukin-4-dependent isotype switching to IgE isotypes
          receptor binding
          cytokine activity
          tumor necrosis factor receptor binding
          tumor necrosis factor receptor superfamily binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 ENST00000367716.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 ENST00000367716.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 ENST00000367716.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 ENST00000367716.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 ENST00000367716.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 ENST00000367716.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 ENST00000367716.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 ENST00000367716.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 ENST00000367716.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 ENST00000367716.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 ENST00000367716.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 ENST00000437190.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 ENST00000437190.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 ENST00000437190.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 ENST00000437190.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 ENST00000437190.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 ENST00000437190.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 NR_037845.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 NR_037845.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 NR_037845.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 NR_037845.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 NR_125960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 NR_125960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 NR_125960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 NR_125960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 NR_125960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 NR_125960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 NR_125960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 NR_125960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 NR_125960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 NR_125960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016432.hg.1 NR_125960.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016431.hg.1 ENST00000281834.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016431.hg.1 ENST00000281834.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016431.hg.1 ENST00000281834.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016431.hg.1 ENST00000281834.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016431.hg.1 ENST00000281834.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016431.hg.1 ENST00000367718.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016431.hg.1 ENST00000367718.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016431.hg.1 ENST00000367718.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016431.hg.1 ENST00000367718.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016431.hg.1 ENST00000488053.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016431.hg.1 ENST00000488053.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016431.hg.1 ENST00000488053.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016431.hg.1 ENST00000488053.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016431.hg.1 NM_001297562.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016431.hg.1 NM_001297562.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016431.hg.1 NM_001297562.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016431.hg.1 NM_001297562.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016431.hg.1 NM_003326.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016431.hg.1 NM_003326.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016431.hg.1 NM_003326.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016431.hg.1 NM_003326.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016431.hg.1 NM_003326.4 Refseq
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 ENST00000367716.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 ENST00000367716.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 ENST00000367716.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 ENST00000367716.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 ENST00000367716.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 ENST00000367716.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 ENST00000367716.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 ENST00000367716.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 ENST00000367716.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 ENST00000367716.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 ENST00000367716.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 ENST00000437190.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 ENST00000437190.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 ENST00000437190.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 ENST00000437190.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 ENST00000437190.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 ENST00000437190.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 NR_037845.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 NR_037845.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 NR_037845.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 NR_037845.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 NR_125960.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 NR_125960.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 NR_125960.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 NR_125960.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 NR_125960.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 NR_125960.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 NR_125960.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 NR_125960.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 NR_125960.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 NR_125960.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016432.hg.1 NR_125960.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0