Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00182558_m1
          See other TRADD GE Assays ›
          Gene Symbol:
          TRADD
          Gene Name:
          TNFRSF1A associated via death domain
          Gene Aliases:
          Hs.89862
          Chromosome Location:
          Chr.16: 67154185 - 67159909 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00182558_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TRADD
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001323552.1 NP_001310481.1 2 - 3 245 92
          NM_003789.3 NP_003780.1 2 - 3 232 92
          XM_017023815.1 XP_016879304.1 2 - 3 298 92
          GenBank mRNA AK315654.1 - 2 - 3 625 92
          BC004491.2 - 2 - 3 209 3689007 92
          BG717460.1 - 2 - 3 227 92
          BT006934.1 - 1 - 2 152 92
          L41690.1 - 2 - 3 200 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1600010616.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC16001180.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Homo sapiens Human NFkB Pathway Human Apoptosis Human TNFR1 Pathway Human TNF Superfamily Pathway HUMAN CELLULAR APOPTOSIS PATHWAY Inventoried Human_Nfkb Human_Apoptosis Human_TNFR1_Pathway Human_TNF_Superfamily_Pathway Human_Cellular_Apoptosis_Pathway Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TRADD

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          signal transduction
          I-kappaB kinase/NF-kappaB signaling
          extrinsic apoptotic signaling pathway via death domain receptors
          regulation of tumor necrosis factor-mediated signaling pathway
          regulation of necrotic cell death
          tumor necrosis factor-mediated signaling pathway
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of NF-kappaB transcription factor activity
          protein heterooligomerization
          positive regulation of hair follicle development
          death-inducing signaling complex assembly
          extrinsic apoptotic signaling pathway
          activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
          regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          signal transducer activity
          tumor necrosis factor receptor binding
          protein binding
          kinase binding
          protein complex binding
          identical protein binding
          binding, bridging
          death domain binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000345057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000345057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000345057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000345057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000345057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000345057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000345057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000345057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000345057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000345057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000486556.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000486556.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000486556.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000486556.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000486556.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000486556.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000486556.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000486556.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000486556.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000486556.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000563348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000563348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000563348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000563348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000566104.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000566104.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000566104.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000566247.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000566247.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000566247.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 ENST00000566247.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 NM_003789.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 NM_003789.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 NM_003789.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 NM_003789.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 NM_003789.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 NM_003789.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 NM_003789.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 NM_003789.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010616.hg.1 NM_003789.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16016391.hg.1 ENST00000345057.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16016391.hg.1 NM_003789.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16016391.hg.1 TC16001180.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000345057.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000345057.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000345057.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000345057.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000345057.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000345057.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000345057.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000345057.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000345057.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000345057.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000486556.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000486556.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000486556.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000486556.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000486556.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000486556.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000486556.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000486556.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000486556.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000486556.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000563348.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000563348.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000563348.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000563348.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000566104.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000566104.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000566104.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000566247.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000566247.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000566247.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 ENST00000566247.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 NM_003789.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 NM_003789.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 NM_003789.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 NM_003789.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 NM_003789.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 NM_003789.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 NM_003789.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 NM_003789.3 Refseq
          Affymetrix Human Clariom D Assay TC1600010616.hg.1 NM_003789.3 Refseq
          Affymetrix Human Clariom D Assay PSR1600181664.hg.1 ENST00000345057.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1600181664.hg.1 ENST00000486556.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600181664.hg.1 ENST00000563348.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600181664.hg.1 NM_003789.3 Refseq
          Affymetrix Human Clariom D Assay PSR1600181664.hg.1 ENST00000345057.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1600181664.hg.1 ENST00000486556.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600181664.hg.1 ENST00000563348.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600181664.hg.1 NM_003789.3 Refseq
          Affymetrix Human Clariom D Assay PSR1600181664.hg.1 TC1600010616.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0