Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00193642_m1
          See other PPP1R3C GE Assays ›
          Gene Symbol:
          PPP1R3C
          Gene Name:
          protein phosphatase 1 regulatory subunit 3C
          Gene Aliases:
          PPP1R5
          Chromosome Location:
          Chr.10: 91628440 - 91633101 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00193642_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PPP1R3C
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005398.5 NP_005389.1 1 - 2 149 61
          GenBank mRNA AK299392.1 - 1 - 2 50 61
          AK299398.1 - 1 - 2 119 61
          AK313149.1 - 1 - 2 119 61
          BC012625.1 - 1 - 2 74 4245774 61
          BP202053.1 - 1 - 2 149 61
          BX537399.1 - 1 - 2 150 61
          Y18207.1 - 1 - 2 108 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000011411.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10001522.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK299392.1, AK299398.1, AK313149.1, BC012625.1, BX537399.1, NM_005398.5, Y18207.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Human Erythropoietin Pathway 1700 Inventoried Human_Erythropoietin_Pathway Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PPP1R3C

          Gene Ontology Categories:

          Function(s) Process(es)

          glycogen metabolic process
          glycogen biosynthetic process
          protein dephosphorylation
          regulation of catalytic activity
          protein serine/threonine phosphatase activity
          protein binding
          protein phosphatase regulator activity
          protein phosphatase binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10019133.hg.1 ENST00000238994.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10019133.hg.1 ENST00000438999.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10019133.hg.1 NM_005398.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10019133.hg.1 TC10001522.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10019137.hg.1 ENST00000238994.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10019137.hg.1 ENST00000500094.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10019137.hg.1 NM_005398.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10019137.hg.1 TC10001522.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000011411.hg.1 ENST00000238994.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011411.hg.1 ENST00000238994.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011411.hg.1 ENST00000238994.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011411.hg.1 ENST00000238994.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011411.hg.1 NM_005398.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011411.hg.1 NM_005398.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011411.hg.1 NM_005398.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011411.hg.1 NM_005398.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011411.hg.1 NM_005398.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10019129.hg.1 ENST00000238994.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10019129.hg.1 ENST00000500094.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10019129.hg.1 NM_005398.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10019129.hg.1 TC10001522.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10019131.hg.1 ENST00000238994.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10019131.hg.1 ENST00000438999.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10019131.hg.1 NM_005398.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10019131.hg.1 TC10001522.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10019134.hg.1 ENST00000238994.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10019134.hg.1 ENST00000438999.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10019134.hg.1 ENST00000500094.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10019134.hg.1 NM_005398.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10019134.hg.1 TC10001522.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10019139.hg.1 NM_005398.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10019139.hg.1 TC10001522.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10019135.hg.1 ENST00000238994.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10019135.hg.1 ENST00000438999.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10019135.hg.1 ENST00000500094.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10019135.hg.1 NM_005398.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10019135.hg.1 TC10001522.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10019136.hg.1 ENST00000238994.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10019136.hg.1 ENST00000500094.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10019136.hg.1 NM_005398.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10019136.hg.1 TC10001522.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10019128.hg.1 ENST00000238994.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10019128.hg.1 ENST00000438999.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10019128.hg.1 ENST00000500094.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10019128.hg.1 NM_005398.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10019128.hg.1 TC10001522.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1000177478.hg.1 ENST00000238994.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000177478.hg.1 NM_005398.6 Refseq
          Affymetrix Human Clariom D Assay PSR1000177478.hg.1 TC1000011411.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000177478.hg.1 NM_005398.6 Refseq
          Affymetrix Human Clariom D Assay PSR1000177478.hg.1 ENST00000238994.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000177479.hg.1 ENST00000238994.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000177479.hg.1 NM_005398.6 Refseq
          Affymetrix Human Clariom D Assay PSR1000177479.hg.1 TC1000011411.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000177479.hg.1 ENST00000238994.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000177479.hg.1 NM_005398.6 Refseq
          Affymetrix Human Clariom D Assay TC1000011411.hg.1 ENST00000238994.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000011411.hg.1 ENST00000238994.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000011411.hg.1 ENST00000238994.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000011411.hg.1 ENST00000238994.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000011411.hg.1 NM_005398.6 Refseq
          Affymetrix Human Clariom D Assay TC1000011411.hg.1 NM_005398.6 Refseq
          Affymetrix Human Clariom D Assay TC1000011411.hg.1 NM_005398.6 Refseq
          Affymetrix Human Clariom D Assay TC1000011411.hg.1 NM_005398.6 Refseq
          Affymetrix Human Clariom D Assay TC1000011411.hg.1 NM_005398.6 Refseq
          Affymetrix Human Clariom D Assay PSR1000177482.hg.1 ENST00000238994.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000177482.hg.1 NM_005398.6 Refseq
          Affymetrix Human Clariom D Assay PSR1000177482.hg.1 TC1000011411.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000177482.hg.1 ENST00000238994.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000177482.hg.1 NM_005398.6 Refseq
          Affymetrix Human Clariom D Assay PSR1000177483.hg.1 NM_005398.6 Refseq
          Affymetrix Human Clariom D Assay PSR1000177483.hg.1 TC1000011411.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000177483.hg.1 NM_005398.6 Refseq
          Affymetrix Human Clariom D Assay PSR1000177481.hg.1 ENST00000238994.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000177481.hg.1 NM_005398.6 Refseq
          Affymetrix Human Clariom D Assay PSR1000177481.hg.1 TC1000011411.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000177481.hg.1 ENST00000238994.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000177481.hg.1 NM_005398.6 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 157725 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0