Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00199607_m1
          See other RPH3AL GE Assays ›
          Gene Symbol:
          RPH3AL
          Gene Name:
          rabphilin 3A-like (without C2 domains)
          Gene Aliases:
          NOC2
          Chromosome Location:
          Chr.17: 212389 - 386254 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00199607_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RPH3AL
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001190411.1 NP_001177340.1 5 - 6 606 140
          NM_006987.3 NP_008918.1 6 - 7 782 140
          XM_011535367.2 XP_011533669.1 5 - 6 533 140
          XM_017025368.1 XP_016880857.1 5 - 6 651 140
          XM_017025370.1 XP_016880859.1 7 - 8 1090 140
          GenBank mRNA AF129812.1 - 5 - 6 687 140
          AK000469.1 - 6 - 7 725 140
          BC093776.1 - 4 - 5 476 7939621 140
          BC113413.1 - 4 - 5 476 8322465 140
          BC143311.1 - 4 - 5 476 140
          BU197069.1 - 5 - 6 549 140

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as HTA transcript cluster(s) TC17000963.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RPH3AL

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          exocytosis
          vesicle fusion
          response to xenobiotic stimulus
          regulation of calcium ion-dependent exocytosis
          glucose homeostasis
          negative regulation of G protein-coupled receptor signaling pathway
          calcium ion-regulated exocytosis of neurotransmitter
          positive regulation of protein secretion
          calcium ion binding
          protein binding
          calcium-dependent phospholipid binding
          cytoskeletal protein binding
          syntaxin binding
          LIM domain binding
          clathrin binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR17012724.hg.1 ENST00000323434.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17012724.hg.1 ENST00000331302.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17012724.hg.1 ENST00000536489.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17012724.hg.1 NM_001190411.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17012724.hg.1 NM_001190412.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17012724.hg.1 NM_001190413.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17012724.hg.1 NM_006987.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17012724.hg.1 TC17000963.hg.1 HTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0