Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00209790_m1
          See other PANX1 GE Assays ›
          Gene Symbol:
          PANX1
          Gene Name:
          pannexin 1
          Gene Aliases:
          MRS1, PX1, UNQ2529
          Chromosome Location:
          Chr.11: 94128928 - 94185596 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00209790_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PANX1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_015368.3 NP_056183.2 2 - 3 708 65
          GenBank mRNA AF398509.1 - 2 - 3 706 65
          AK074897.1 - 2 - 3 653 65
          AK313074.1 - 2 - 3 678 65
          AY359023.1 - 2 - 3 469 65
          BC016931.1 - 2 - 3 703 4390851 65
          FM201789.1 - 2 - 3 323 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100008797.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11000916.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PANX1

          Gene Ontology Categories:

          Function(s) Process(es)

          response to ischemia
          cation transport
          calcium ion transport
          cell-cell signaling
          response to ATP
          protein hexamerization
          calcium ion transmembrane transport
          protease binding
          signaling receptor binding
          calcium channel activity
          leak channel activity
          transmembrane transporter binding
          protein heterodimerization activity
          actin filament binding
          gap junction hemi-channel activity
          scaffold protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR11011486.hg.1 ENST00000227638.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11011486.hg.1 ENST00000436171.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11011486.hg.1 NM_015368.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11011486.hg.1 TC11000916.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100008797.hg.1 ENST00000227638.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008797.hg.1 ENST00000227638.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008797.hg.1 ENST00000227638.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008797.hg.1 ENST00000227638.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008797.hg.1 ENST00000227638.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008797.hg.1 ENST00000227638.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008797.hg.1 ENST00000227638.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008797.hg.1 ENST00000227638.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008797.hg.1 ENST00000436171.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008797.hg.1 ENST00000436171.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008797.hg.1 ENST00000436171.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008797.hg.1 ENST00000436171.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008797.hg.1 ENST00000436171.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008797.hg.1 ENST00000436171.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008797.hg.1 ENST00000436171.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008797.hg.1 ENST00000436171.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008797.hg.1 NM_015368.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008797.hg.1 NM_015368.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008797.hg.1 NM_015368.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008797.hg.1 NM_015368.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008797.hg.1 NM_015368.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008797.hg.1 NM_015368.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008797.hg.1 NM_015368.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008797.hg.1 NM_015368.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100164979.hg.1 ENST00000227638.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100164979.hg.1 ENST00000436171.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100164979.hg.1 NM_015368.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100164979.hg.1 TC1100008797.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100164979.hg.1 NM_015368.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100164979.hg.1 ENST00000227638.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100164979.hg.1 ENST00000436171.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008797.hg.1 ENST00000227638.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008797.hg.1 ENST00000227638.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008797.hg.1 ENST00000227638.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008797.hg.1 ENST00000227638.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008797.hg.1 ENST00000227638.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008797.hg.1 ENST00000227638.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008797.hg.1 ENST00000227638.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008797.hg.1 ENST00000227638.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100008797.hg.1 ENST00000436171.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008797.hg.1 ENST00000436171.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008797.hg.1 ENST00000436171.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008797.hg.1 ENST00000436171.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008797.hg.1 ENST00000436171.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008797.hg.1 ENST00000436171.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008797.hg.1 ENST00000436171.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008797.hg.1 ENST00000436171.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008797.hg.1 NM_015368.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008797.hg.1 NM_015368.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008797.hg.1 NM_015368.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008797.hg.1 NM_015368.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008797.hg.1 NM_015368.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008797.hg.1 NM_015368.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008797.hg.1 NM_015368.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008797.hg.1 NM_015368.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 199931 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc