Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs00222461_m1
See other DTNB GE Assays ›
Gene Symbol:
DTNB
Gene Name:
dystrobrevin beta
Gene Aliases:
-
Chromosome Location:
Chr.2: 25377220 - 25673647 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs00222461_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Inventoried
Catalog #
4331182
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: DTNB
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001320934.1
NP_001307863.1
15 - 16
1749
66
NM_001320936.1
NP_001307865.1
16 - 17
1839
66
NM_021907.4
NP_068707.1
16 - 17
1839
66
NM_033147.3
NP_149159.2
16 - 17
1839
66
NM_033148.3
NP_149160.1
15 - 16
1749
66
NM_183360.2
NP_899204.1
16 - 17
1839
66
XM_011532671.1
XP_011530973.1
16 - 17
1774
66
XM_011532672.2
XP_011530974.1
16 - 17
1774
66
XM_011532674.1
XP_011530976.1
16 - 17
1774
66
XM_011532676.1
XP_011530978.1
15 - 16
1684
66
XM_011532677.2
XP_011530979.1
15 - 16
2230
66
XM_011532680.2
XP_011530982.1
15 - 16
1695
66
XM_011532681.2
XP_011530983.1
15 - 16
1670
66
XM_017003532.1
XP_016859021.1
15 - 16
1684
66
XM_017003533.1
XP_016859022.1
15 - 16
1666
66
XM_017003537.1
XP_016859026.1
12 - 13
2127
66
XM_017003539.1
XP_016859028.1
14 - 15
1541
66
XM_017003540.1
XP_016859029.1
14 - 15
1580
66
XM_017003541.1
XP_016859030.1
14 - 15
1564
66
XM_017003542.1
XP_016859031.1
14 - 15
1536
66
XM_017003543.1
XP_016859032.1
13 - 14
1430
66
XM_017003544.1
XP_016859033.1
14 - 15
1518
66
XM_017003545.1
XP_016859034.1
13 - 14
1412
66
GenBank mRNA
AF022728.1
-
16 - 17
1737
66
AF070567.1
-
10 - 11
981
66
AK094494.1
-
15 - 16
1557
66
BC049366.1
-
16 - 17
1830
4817552
66
Y12712.1
-
15 - 16
1669
66
Y15722.1
-
15 - 16
1576
66
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length less than or equal to 70
Mammalian Gene Collection
Homo sapiens
Inventoried
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene DTNB
Gene Ontology Categories:
Process(es)
protein binding
zinc ion binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0