Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00231279_m1
          See other REL GE Assays ›
          Gene Symbol:
          REL
          Gene Name:
          REL proto-oncogene, NF-kB subunit
          Gene Aliases:
          C-Rel
          Chromosome Location:
          Chr.2: 60881495 - 60928171 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00231279_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: REL
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001291746.1 NP_001278675.1 1 - 2 356 134
          NM_002908.3 NP_002899.1 1 - 2 356 134
          XM_017004627.1 XP_016860116.1 1 - 2 277 134
          GenBank mRNA AK303926.1 - 1 - 2 277 134
          BC117191.1 - 1 - 2 195 40125742 134
          BC143885.1 - 1 - 2 197 9052401 134
          CN414487.1 - 1 - 2 235 134
          HY123671.1 - 1 - 2 355 134
          X75042.1 - 1 - 2 187 134

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200016452.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02000348.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK303926.1,BC117191.1,BC143885.1,NM_001291746.1,NM_002908.3,X75042.1,XM_017004627.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Immune Tolerance Network Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene REL

          Panther Classification:

          Molecular Function -

          Rel homology transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          inflammatory response
          I-kappaB kinase/NF-kappaB signaling
          negative regulation of gene expression
          negative regulation of interferon-beta production
          response to cytokine
          NIK/NF-kappaB signaling
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          innate immune response
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of neuron death
          RNA polymerase II distal enhancer sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02005171.hg.1 TC02000348.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02005171.hg.1 ENST00000295025.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02005171.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02005171.hg.1 NM_002908.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02005168.hg.1 TC02000348.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02005168.hg.1 ENST00000295025.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02005168.hg.1 NM_002908.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02005170.hg.1 TC02000348.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02005170.hg.1 ENST00000295025.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02005170.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02005170.hg.1 NM_002908.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02005172.hg.1 TC02000348.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02005172.hg.1 ENST00000295025.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02005172.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02005172.hg.1 NM_002908.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000589496.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 ENST00000589496.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02005173.hg.1 TC02000348.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02005173.hg.1 ENST00000295025.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02005173.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02005173.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200221884.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Clariom D Assay PSR0200221884.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200221884.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200221884.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Clariom D Assay PSR0200221884.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200221884.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200221884.hg.1 TC0200016452.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200221883.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200221883.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200221883.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200221883.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200221883.hg.1 TC0200016452.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000589496.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 ENST00000589496.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay TC0200016452.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200221886.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Clariom D Assay PSR0200221886.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0200221886.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200221886.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200221886.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Clariom D Assay PSR0200221886.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0200221886.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200221886.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200221886.hg.1 TC0200016452.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200221891.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Clariom D Assay PSR0200221891.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0200221891.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200221891.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200221891.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Clariom D Assay PSR0200221891.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0200221891.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200221891.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200221891.hg.1 TC0200016452.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200221889.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Clariom D Assay PSR0200221889.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0200221889.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200221889.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200221889.hg.1 ENST00000295025.12 Ensembl
          Affymetrix Human Clariom D Assay PSR0200221889.hg.1 ENST00000394479.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0200221889.hg.1 NM_001291746.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200221889.hg.1 NM_002908.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200221889.hg.1 TC0200016452.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6f7b98477d-hxtzg:80/100.66.130.62:80.
          git-commit: 5163bf559e7ad0ebe9326a88094353fc6888890d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline