Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00231692_m1
          See other RUNX2 GE Assays ›
          Gene Symbol:
          RUNX2
          Gene Name:
          runt related transcription factor 2
          Gene Aliases:
          AML3, CBF-alpha-1, CBFA1, CCD, CCD1, CLCD, OSF-2, OSF2, PEA2aA, PEBP2aA
          Chromosome Location:
          Chr.6: 45328142 - 45664032 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00231692_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RUNX2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001015051.3 NP_001015051.3 5 - 6 900 116
          NM_001024630.3 NP_001019801.3 5 - 6 900 116
          NM_001278478.1 NP_001265407.1 3 - 4 648 116
          XM_011514960.2 XP_011513262.1 5 - 6 1075 116
          XM_011514961.2 XP_011513263.1 5 - 6 1075 116
          XM_011514962.2 XP_011513264.1 5 - 6 1075 116
          XM_011514964.2 XP_011513266.1 5 - 6 1075 116
          XM_011514965.2 XP_011513267.1 5 - 6 1075 116
          XM_011514966.2 XP_011513268.1 3 - 4 1718 116
          XM_017011391.1 XP_016866880.1 5 - 6 1110 116
          XM_017011392.1 XP_016866881.1 3 - 4 651 116
          XM_017011393.1 XP_016866882.1 5 - 6 1075 116
          GenBank mRNA BC108919.1 - 3 - 4 630 116
          BC108920.1 - 3 - 4 634 116
          L40992.1 - 3 - 4 513 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600008146.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000621.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Human Osteogenesis Human Stem Cell Pluripotency Inventoried Human_Osteogenesis Human_Stem_Cell Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RUNX2

          Panther Classification:

          Molecular Function -

          Runt transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          osteoblast differentiation
          endochondral ossification
          osteoblast fate commitment
          chondrocyte differentiation
          chondrocyte development
          osteoblast development
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          positive regulation of cell proliferation
          hemopoiesis
          neuron differentiation
          T cell differentiation
          BMP signaling pathway
          positive regulation of chondrocyte differentiation
          embryonic forelimb morphogenesis
          regulation of fibroblast growth factor receptor signaling pathway
          odontogenesis of dentin-containing tooth
          regulation of odontogenesis of dentin-containing tooth
          cellular response to fibroblast growth factor stimulus
          regulation of cell differentiation
          positive regulation of osteoblast differentiation
          negative regulation of smoothened signaling pathway
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          cell maturation
          embryonic cranial skeleton morphogenesis
          stem cell differentiation
          cellular response to BMP stimulus
          positive regulation of transcription from RNA polymerase II promoter involved in cellular response to chemical stimulus
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          ATP binding
          protein domain specific binding
          histone deacetylase binding
          bHLH transcription factor binding
          repressing transcription factor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000359524.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000359524.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000359524.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000359524.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000359524.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000359524.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000359524.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000359524.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000359524.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000359524.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000359524.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371436.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371436.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371436.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371436.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371436.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371436.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371436.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371436.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371436.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000371438.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000465038.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000465038.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000465038.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000465038.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000465038.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000465038.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000465038.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000465038.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000465038.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000465038.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000465038.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000465038.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000473041.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000473041.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000478660.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000478660.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000478660.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000478660.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000478660.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000478660.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000478660.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000478660.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483243.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483243.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483243.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483243.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483243.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483243.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483377.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483377.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483377.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483377.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483377.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483377.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483377.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483377.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483377.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483377.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000483377.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000576263.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000576263.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000576263.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000576263.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000576263.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000576263.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000576263.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000576263.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000625924.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000625924.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000625924.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000625924.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000625924.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000625924.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 ENST00000625924.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001015051.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001024630.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001278478.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001278478.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001278478.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001278478.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001278478.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001278478.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001278478.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001278478.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NM_001278478.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NR_103532.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NR_103532.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NR_103532.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NR_103532.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NR_103532.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NR_103532.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NR_103533.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NR_103533.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NR_103533.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NR_103533.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008146.hg.1 NR_103533.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0