Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00233552_m1
          See other CD38 GE Assays ›
          Gene Symbol:
          CD38
          Gene Name:
          CD38 molecule
          Gene Aliases:
          ADPRC 1, ADPRC1
          Chromosome Location:
          Chr.4: 15778265 - 15853243 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00233552_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CD38
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001775.3 NP_001766.2 2 - 3 514 119
          GenBank mRNA AK313268.1 - 2 - 3 451 119
          BC007964.2 - 2 - 3 412 4309086 119
          CD688069.1 - 2 - 3 505 119
          D84276.1 - 2 - 3 467 119
          M34461.1 - 2 - 3 433 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400006951.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC04000145.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Immune Tolerance Network Mammalian Gene Collection Homo sapiens Human Immune Response 1700 Inventoried Human_Immune Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CD38

          Panther Classification:

          Molecular Function -

          cyclase lyase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          signal transduction
          positive regulation of cytosolic calcium ion concentration
          female pregnancy
          positive regulation of cell growth
          positive regulation of B cell proliferation
          positive regulation of insulin secretion
          response to estradiol
          response to retinoic acid
          response to progesterone
          response to hydroperoxide
          response to drug
          negative regulation of apoptotic process
          negative regulation of bone resorption
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of vasoconstriction
          B cell receptor signaling pathway
          long term synaptic depression
          response to interleukin-1
          apoptotic signaling pathway
          NAD+ nucleosidase activity
          transferase activity
          phosphorus-oxygen lyase activity
          NAD(P)+ nucleosidase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR04002316.hg.1 TC04000145.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04002316.hg.1 ENST00000226279.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04002316.hg.1 NM_001775.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000506191.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000506191.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000506191.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000506191.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000506191.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000506191.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000510674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000510674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000510674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000510674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000510674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000510674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000511430.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000511430.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000511430.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000511430.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000511430.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000511430.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 ENST00000511430.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400147790.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0400147790.hg.1 ENST00000510674.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400147790.hg.1 ENST00000511430.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400147790.hg.1 NM_001775.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400147790.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0400147790.hg.1 ENST00000510674.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400147790.hg.1 ENST00000511430.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400147790.hg.1 NM_001775.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400147790.hg.1 TC0400006951.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000226279.7 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000502843.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000506191.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000506191.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000506191.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000506191.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000506191.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000506191.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000510674.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000510674.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000510674.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000510674.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000510674.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000510674.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000511430.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000511430.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000511430.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000511430.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000511430.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000511430.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 ENST00000511430.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 NM_001775.3 Refseq
          Affymetrix Human Clariom D Assay TC0400006951.hg.1 NM_001775.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 162896 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0