Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00234646_m1
          See other CCL13 GE Assays ›
          Gene Symbol:
          CCL13
          Gene Name:
          C-C motif chemokine ligand 13
          Gene Aliases:
          CKb10, MCP-4, NCC-1, NCC1, SCYA13, SCYL1
          Chromosome Location:
          Chr.17: 34356452 - 34358610 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00234646_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CCL13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005408.2 NP_005399.1 1 - 2 154 74
          GenBank mRNA AJ001634.1 - 1 - 2 121 74
          BC008621.1 - 1 - 2 117 4184250 74
          BT007385.1 - 1 - 2 79 74
          CR450337.1 - 1 - 2 79 74
          U46767.1 - 1 - 2 112 74
          U59808.1 - 1 - 2 154 74
          X98306.1 - 1 - 2 108 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700007561.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17000387.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Immune Tolerance Network Mammalian Gene Collection Recommended Assay Homo sapiens Human Chemokine Panel Inventoried Human_Chemokine Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CCL13

          Panther Classification:

          Molecular Function -

          cytokine intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          monocyte chemotaxis
          cellular calcium ion homeostasis
          chemotaxis
          inflammatory response
          cytoskeleton organization
          signal transduction
          G-protein coupled receptor signaling pathway
          cell-cell signaling
          regulation of cell shape
          neutrophil chemotaxis
          positive regulation of GTPase activity
          eosinophil chemotaxis
          lymphocyte chemotaxis
          positive regulation of inflammatory response
          chemokine-mediated signaling pathway
          positive regulation of ERK1 and ERK2 cascade
          cellular response to interferon-gamma
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          receptor binding
          chemokine activity
          CCR chemokine receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR17004895.hg.1 ENST00000225844.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17004895.hg.1 NM_005408.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17004895.hg.1 TC17000387.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1700007561.hg.1 ENST00000225844.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007561.hg.1 ENST00000225844.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007561.hg.1 ENST00000225844.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007561.hg.1 ENST00000225844.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007561.hg.1 ENST00000225844.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007561.hg.1 ENST00000225844.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007561.hg.1 ENST00000577681.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007561.hg.1 ENST00000577681.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007561.hg.1 ENST00000577681.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007561.hg.1 NM_005408.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007561.hg.1 NM_005408.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007561.hg.1 NM_005408.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007561.hg.1 NM_005408.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007561.hg.1 NM_005408.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007561.hg.1 NM_005408.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17004894.hg.1 ENST00000225844.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17004894.hg.1 NM_005408.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17004894.hg.1 TC17000387.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17004893.hg.1 ENST00000225844.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17004893.hg.1 NM_005408.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17004893.hg.1 TC17000387.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1700007561.hg.1 ENST00000225844.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700007561.hg.1 ENST00000225844.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700007561.hg.1 ENST00000225844.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700007561.hg.1 ENST00000225844.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700007561.hg.1 ENST00000225844.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700007561.hg.1 ENST00000225844.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700007561.hg.1 ENST00000577681.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007561.hg.1 ENST00000577681.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007561.hg.1 ENST00000577681.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007561.hg.1 NM_005408.2 Refseq
          Affymetrix Human Clariom D Assay TC1700007561.hg.1 NM_005408.2 Refseq
          Affymetrix Human Clariom D Assay TC1700007561.hg.1 NM_005408.2 Refseq
          Affymetrix Human Clariom D Assay TC1700007561.hg.1 NM_005408.2 Refseq
          Affymetrix Human Clariom D Assay TC1700007561.hg.1 NM_005408.2 Refseq
          Affymetrix Human Clariom D Assay TC1700007561.hg.1 NM_005408.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700154410.hg.1 ENST00000225844.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1700154410.hg.1 NM_005408.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700154410.hg.1 ENST00000225844.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1700154410.hg.1 NM_005408.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700154410.hg.1 TC1700007561.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700154411.hg.1 ENST00000225844.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1700154411.hg.1 NM_005408.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700154411.hg.1 ENST00000225844.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1700154411.hg.1 NM_005408.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700154411.hg.1 TC1700007561.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700154412.hg.1 ENST00000225844.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1700154412.hg.1 NM_005408.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700154412.hg.1 ENST00000225844.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1700154412.hg.1 NM_005408.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700154412.hg.1 TC1700007561.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700154414.hg.1 ENST00000225844.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1700154414.hg.1 ENST00000577681.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700154414.hg.1 NM_005408.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700154414.hg.1 ENST00000225844.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1700154414.hg.1 ENST00000577681.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700154414.hg.1 NM_005408.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700154414.hg.1 TC1700007561.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0