Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00236966_m1
          See other CXCL2 GE Assays ›
          Gene Symbol:
          CXCL2
          Gene Name:
          C-X-C motif chemokine ligand 2
          Gene Aliases:
          CINC-2a, GRO2, GROb, MGSA-b, MIP-2a, MIP2, MIP2A, SCYB2
          Chromosome Location:
          Chr.4: 74097035 - 74099280 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00236966_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CXCL2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002089.3 NP_002080.1 3 - 4 465 68
          GenBank mRNA AF043340.1 - 2 - 3 203 68
          BC015753.1 - 3 - 4 369 4875789 68
          BX334969.2 - 3 - 4 475 68
          M36820.1 - 3 - 4 379 68
          M57731.1 - 3 - 4 374 68
          X53799.1 - 3 - 4 347 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400011018.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC04001286.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Human Chemokine Panel 1700 Inventoried Human_Chemokine Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CXCL2

          Panther Classification:

          Molecular Function -

          chemokine

          Gene Ontology Categories:

          Function(s) Process(es)

          response to molecule of bacterial origin
          chemotaxis
          inflammatory response
          immune response
          G-protein coupled receptor signaling pathway
          response to lipopolysaccharide
          regulation of cell proliferation
          cell chemotaxis
          chemokine-mediated signaling pathway
          positive regulation of neutrophil chemotaxis
          protein binding
          chemokine activity
          CXCR chemokine receptor binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR04017574.hg.1 TC04001286.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04017574.hg.1 ENST00000296031.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04017574.hg.1 ENST00000508487.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04017574.hg.1 NM_002089.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04017578.hg.1 TC04001286.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04017578.hg.1 ENST00000296031.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04017578.hg.1 ENST00000508487.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04017578.hg.1 NM_002089.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 ENST00000296031.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 ENST00000296031.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 ENST00000296031.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 ENST00000296031.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 ENST00000296031.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 ENST00000296031.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 ENST00000508487.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 ENST00000508487.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 ENST00000508487.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 ENST00000508487.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 ENST00000508487.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 ENST00000508487.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 ENST00000508487.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 ENST00000510048.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 ENST00000510048.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 ENST00000510048.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 ENST00000510048.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 ENST00000510048.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 ENST00000510048.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 NM_002089.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 NM_002089.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 NM_002089.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 NM_002089.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 NM_002089.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 NM_002089.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011018.hg.1 NM_002089.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04017580.hg.1 TC04001286.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04017580.hg.1 ENST00000296031.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04017580.hg.1 ENST00000508487.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04017580.hg.1 NM_002089.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04017576.hg.1 TC04001286.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04017576.hg.1 ENST00000296031.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04017576.hg.1 ENST00000508487.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04017576.hg.1 NM_002089.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04017583.hg.1 TC04001286.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04017583.hg.1 ENST00000508487.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04017583.hg.1 NM_002089.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011011.hg.1 ENST00000502804.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011011.hg.1 ENST00000502804.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174919.hg.1 ENST00000296031.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174919.hg.1 ENST00000508487.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174919.hg.1 ENST00000510048.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174919.hg.1 NM_002089.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400174919.hg.1 ENST00000296031.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174919.hg.1 ENST00000508487.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174919.hg.1 ENST00000510048.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174919.hg.1 NM_002089.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400174919.hg.1 TC0400011018.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400174922.hg.1 ENST00000508487.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174922.hg.1 NM_002089.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400174922.hg.1 ENST00000508487.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174922.hg.1 NM_002089.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400174922.hg.1 TC0400011018.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400174914.hg.1 ENST00000296031.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174914.hg.1 ENST00000508487.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174914.hg.1 ENST00000510048.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174914.hg.1 NM_002089.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400174914.hg.1 ENST00000296031.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174914.hg.1 ENST00000508487.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174914.hg.1 ENST00000510048.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174914.hg.1 NM_002089.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400174914.hg.1 TC0400011018.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 ENST00000296031.4 Ensembl
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 ENST00000296031.4 Ensembl
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 ENST00000296031.4 Ensembl
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 ENST00000296031.4 Ensembl
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 ENST00000296031.4 Ensembl
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 ENST00000296031.4 Ensembl
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 ENST00000508487.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 ENST00000508487.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 ENST00000508487.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 ENST00000508487.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 ENST00000508487.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 ENST00000508487.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 ENST00000508487.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 ENST00000510048.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 ENST00000510048.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 ENST00000510048.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 ENST00000510048.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 ENST00000510048.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 ENST00000510048.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 NM_002089.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 NM_002089.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 NM_002089.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 NM_002089.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 NM_002089.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 NM_002089.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011018.hg.1 NM_002089.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400174918.hg.1 ENST00000296031.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174918.hg.1 ENST00000508487.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174918.hg.1 ENST00000510048.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174918.hg.1 NM_002089.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400174918.hg.1 ENST00000296031.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174918.hg.1 ENST00000508487.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174918.hg.1 ENST00000510048.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174918.hg.1 NM_002089.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400174918.hg.1 TC0400011018.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400174916.hg.1 ENST00000296031.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174916.hg.1 ENST00000508487.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174916.hg.1 ENST00000510048.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174916.hg.1 NM_002089.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400174916.hg.1 ENST00000296031.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174916.hg.1 ENST00000508487.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174916.hg.1 ENST00000510048.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400174916.hg.1 NM_002089.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400174916.hg.1 TC0400011018.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0400011011.hg.1 ENST00000502804.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011011.hg.1 ENST00000502804.2 Ensembl
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 173408 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0