Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00237035_m1
          See other TRAF3 GE Assays ›
          Gene Symbol:
          TRAF3
          Gene Name:
          TNF receptor associated factor 3
          Gene Aliases:
          CAP-1, CAP1, CD40bp, CRAF1, IIAE5, LAP1
          Chromosome Location:
          Chr.14: 102777479 - 102911500 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00237035_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TRAF3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003300.3 NP_003291.2 5 - 6 783 66
          NM_145725.2 NP_663777.1 6 - 7 922 66
          NM_145726.2 NP_663778.1 6 - 7 922 66
          XM_011537117.2 XP_011535419.1 6 - 7 898 66
          XM_017021617.1 XP_016877106.1 6 - 7 1342 66
          XM_017021618.1 XP_016877107.1 6 - 7 2838 66
          XM_017021619.1 XP_016877108.1 6 - 7 2840 66
          XM_017021620.1 XP_016877109.1 6 - 7 900 66
          GenBank mRNA BC075086.2 - 4 - 5 594 30915387 66
          BC075087.2 - 4 - 5 594 30915599 66
          BX247977.1 - 6 - 7 768 66
          L38509.1 - 5 - 6 705 66
          U15637.1 - 6 - 7 776 66
          U19260.1 - 5 - 6 719 66
          U21092.1 - 5 - 6 786 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1400008362.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC14000786.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Human NFkB Pathway Inventoried Human_Nfkb Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TRAF3

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cytokine production
          toll-like receptor signaling pathway
          apoptotic process
          signal transduction
          Toll signaling pathway
          protein ubiquitination
          regulation of proteolysis
          negative regulation of NF-kappaB transcription factor activity
          negative regulation of type I interferon production
          regulation of interferon-beta production
          tumor necrosis factor-mediated signaling pathway
          TRIF-dependent toll-like receptor signaling pathway
          regulation of apoptotic process
          innate immune response
          regulation of defense response to virus
          ubiquitin-protein transferase activity
          tumor necrosis factor receptor binding
          protein binding
          zinc ion binding
          ligase activity
          protein kinase binding
          protein phosphatase binding
          ubiquitin protein ligase binding
          thioesterase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR14008513.hg.1 ENST00000347662.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14008513.hg.1 ENST00000351691.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14008513.hg.1 ENST00000392745.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14008513.hg.1 ENST00000560371.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14008513.hg.1 NM_003300.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14008513.hg.1 NM_145725.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14008513.hg.1 NM_145726.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14008513.hg.1 TC14000786.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1400159672.hg.1 ENST00000347662.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1400159672.hg.1 ENST00000351691.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1400159672.hg.1 ENST00000392745.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400159672.hg.1 ENST00000558700.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400159672.hg.1 ENST00000560371.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400159672.hg.1 NM_003300.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400159672.hg.1 NM_145725.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400159672.hg.1 NM_145726.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400159672.hg.1 TC1400008362.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400159672.hg.1 ENST00000347662.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1400159672.hg.1 ENST00000351691.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1400159672.hg.1 ENST00000392745.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400159672.hg.1 ENST00000558700.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400159672.hg.1 ENST00000560371.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400159672.hg.1 NM_003300.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400159672.hg.1 NM_145725.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400159672.hg.1 NM_145726.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b7b59cd5b-kqhbv:80/100.66.135.122:80.
          git-commit: 87deb889a5bb48f859ebd9970e87be562fd9396c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0