Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00240887_m1
          See other RET GE Assays ›
          Gene Symbol:
          RET
          Gene Name:
          ret proto-oncogene
          Gene Aliases:
          CDHF12, CDHR16, HSCR1, MEN2A, MEN2B, MTC1, PTC, RET-ELE1, RET51
          Chromosome Location:
          Chr.10: 43077069 - 43130351 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00240887_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RET
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020630.4 NP_065681.1 819 131
          NM_020975.4 NP_066124.1 819 131
          GenBank mRNA AK291807.1 - 689 131
          BC003072.2 - 809 131
          BC004257.1 - 809 3160389 131
          X15262.1 - 778 131

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000007403.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10000268.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Homo sapiens Human Tumor Metastasis Panel Human Protein Kinase Pathways Inventoried Human_Protein_Kinase Human Tumor Metastasis

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RET

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          ureteric bud development
          neural crest cell migration
          embryonic epithelial tube formation
          protein phosphorylation
          homophilic cell adhesion via plasma membrane adhesion molecules
          neuron cell-cell adhesion
          signal transduction
          transmembrane receptor protein tyrosine kinase signaling pathway
          posterior midgut development
          positive regulation of neuron projection development
          positive regulation of neuron maturation
          peptidyl-tyrosine phosphorylation
          regulation of cell adhesion
          positive regulation of cell migration
          membrane protein proteolysis
          positive regulation of cell adhesion mediated by integrin
          ureter maturation
          response to drug
          neuron maturation
          positive regulation of GTPase activity
          positive regulation of cell size
          positive regulation of transcription, DNA-templated
          response to pain
          enteric nervous system development
          regulation of axonogenesis
          retina development in camera-type eye
          innervation
          Peyer's patch morphogenesis
          cellular response to retinoic acid
          positive regulation of metanephric glomerulus development
          lymphocyte migration into lymphoid organs
          positive regulation of serine phosphorylation of STAT3 protein
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          receptor activity
          Ras guanyl-nucleotide exchange factor activity
          calcium ion binding
          protein binding
          ATP binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000479913.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000479913.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000479913.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000498820.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000498820.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000498820.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000498820.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000498820.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000498820.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0