Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00251433_s1
          See other PRO2012 GE Assays ›
          Gene Symbol:
          PRO2012
          Gene Name:
          uncharacterized protein PRO2012
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00251433_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PRO2012
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF116681.2 - 1 194 149

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AF116681.2 transcripts.
          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3821350 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P62793 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P389480 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P62787 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 30030 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 28543 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 42663 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 6279 - -
          SurePrint G3 Human GE Microarray G4851A A_19_P00801188 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3821350 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P62787 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P62793 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P389480 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P62793 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P62787 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P389480 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7924818 - -
          Affymetrix Human Gene 1.1 ST Array 7924817 - -
          Affymetrix HT HG-U133+ PM Array Plate 1553546_PM_at - -
          Human Genome U133 Plus 2.0 Array 1553546_at - -
          Human Genome U133 Plus 2.0 Array 220883_at - -
          Human Genome U133 Plus 2.0 Array 220883_PM_at - -
          Human Genome U133 Plus 2.0 Array 1553546_PM_at - -
          Human Genome U133A 2.0 Array 220883_at - -
          Human Genome U133A 2.0 Array 220883_PM_at - -
          AffymetrixPathwork Diagnostics Pathchip pw587599 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220883_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220883_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553546_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553546_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220883_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553546_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220883_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220883_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553546_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553546_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553546_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220883_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220883_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553546_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553546_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553546_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220883_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220883_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220883_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553546_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220883_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553546_at3 - -
          Human Genome U133 Set 220883_PM_at - -
          Human Genome U133 Set 220883_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 112089 - -
          ABI Human Genome Survey Microarray Version 2.0 221589 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0