Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00271858_m1
          See other IDH1 GE Assays ›
          Gene Symbol:
          IDH1
          Gene Name:
          isocitrate dehydrogenase (NADP(+)) 1, cytosolic
          Gene Aliases:
          HEL-216, HEL-S-26, IDCD, IDH, IDP, IDPC, PICD
          Chromosome Location:
          Chr.2: 208236227 - 208255143 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00271858_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IDH1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001282386.1 NP_001269315.1 2 - 3 189 96
          NM_001282387.1 NP_001269316.1 2 - 3 346 96
          NM_005896.3 NP_005887.2 2 - 3 278 96
          GenBank mRNA AF020038.1 - 2 - 3 232 96
          AF113917.1 - 2 - 3 169 96
          AK312242.1 - 2 - 3 249 96
          AL136702.1 - 2 - 3 210 96
          BC012846.1 - 2 - 3 229 3880331 96
          BC021046.1 - 2 - 3 175 96
          BC093020.1 - 2 - 3 207 30528912 96
          BX537411.1 - 2 - 3 178 96
          DA728868.1 - 2 - 3 278 96
          DR158630.1 - 2 - 3 346 96
          EU794623.1 - 2 - 3 217 96
          FJ224342.1 - 2 - 3 178 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200015578.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02002726.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AF020038.1,AF113917.1,AK312242.1,AL136702.1,BC012846.1,BC093020.1,BX537411.1,EU794623.1,FJ224342.1,NM_001282386.1,NM_001282387.1,NM_005896.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene IDH1

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          glyoxylate cycle
          tricarboxylic acid cycle
          isocitrate metabolic process
          2-oxoglutarate metabolic process
          NADPH regeneration
          glutathione metabolic process
          response to oxidative stress
          female gonad development
          response to steroid hormone
          regulation of phospholipid catabolic process
          regulation of phospholipid biosynthetic process
          cell-cell adhesion
          magnesium ion binding
          isocitrate dehydrogenase (NADP+) activity
          receptor binding
          protein homodimerization activity
          NADP binding
          NAD binding
          (R)-2-hydroxyglutarate dehydrogenase activity
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000417583.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000417583.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000417583.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000417583.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000417583.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000417583.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000417583.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000451391.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000451391.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000451391.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000451391.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000451391.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000451391.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000462386.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000462386.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000462386.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000462386.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000462386.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000462386.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000462386.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000481557.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000481557.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000481557.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000481557.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000481557.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000484575.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000484575.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000484575.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000484575.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000484575.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010739.hg.1 ENST00000440581.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600010739.hg.1 ENST00000440581.1 Ensembl
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 231342 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline