Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00272112_s1
          See other PIN1P1 GE Assays ›
          Gene Symbol:
          PIN1P1
          Gene Name:
          peptidylprolyl cis/trans isomerase, NIMA-interacting 1 pseudogene 1
          Gene Aliases:
          PIN1L
          Chromosome Location:
          Chr.1: 69919322 - 69920317 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00272112_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PIN1P1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_023916.1 1 629 134
          GenBank mRNA U82382.1 - 1 629 134

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P266 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P276493 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P266 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 21402 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 44561 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 6764 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P266 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P266 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00275719 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00275745 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00275701 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304319 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00275749 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304324 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304336 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304349 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304330 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00275725 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304343 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304310 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304317 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304318 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00275726 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304327 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304345 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304323 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304334 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00275751 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304348 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304306 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304351 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00275700 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15070689 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00275732 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304331 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304355 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304313 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304309 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304311 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304339 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304346 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304341 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304342 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304340 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304326 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00275718 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304347 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304347 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00275719 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304334 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304330 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304331 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304348 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304336 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00275725 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304349 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304339 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304340 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304345 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00275751 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304327 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304318 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304326 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00275745 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304319 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304346 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304341 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304310 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304342 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304309 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304343 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304306 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304355 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304311 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304313 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00275749 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00275700 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15070689 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304324 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304317 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00275718 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00275701 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304351 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00275732 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20304323 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00275726 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P276493 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P266 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P276493 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P266 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15070692 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00275722 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00275726 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00275701 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00275728 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00275746 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00275720 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00275702 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00275716 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00275736 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00275730 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00275738 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00275732 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00275747 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15070689 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00275708 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00275731 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00275735 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00275743 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00275714 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00275704 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00275723 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00275718 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00275711 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00275751 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00275707 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7902268 - -
          Affymetrix Human Gene 1.1 ST Array 7902267 - -
          Affymetrix HT HG-U133+ PM Array Plate 207582_PM_at - -
          Affymetrix Human Genome U219 Array 11749082_PM_at - -
          Affymetrix Human Genome U219 Array 11749082_at - -
          Human Genome U133 Plus 2.0 Array 207582_PM_at - -
          Human Genome U133 Plus 2.0 Array 207582_at - -
          Human Genome U133A 2.0 Array 207582_PM_at - -
          Human Genome U133A 2.0 Array 207582_at - -
          AffymetrixPathwork Diagnostics Pathchip pw590583 - -
          Human Genome U95 Set 31717_PM_at - -
          Human Genome U95 Set 31717_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207582_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207582_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207582_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207582_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207582_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207582_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207582_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207582_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207582_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207582_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207582_at5 - -
          Human Genome U133 Set 207582_PM_at - -
          Human Genome U133 Set 207582_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A024245 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A024245 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 159340 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3237693 U82382.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3237693 NR_023916.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3237693 U82382.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3237693 NR_023916.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3237693 U82382.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3237693 NR_023916.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3237693 U82382.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3237693 NR_023916.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0