Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00291414_s1
          See other LOC100129917 GE Assays ›
          Gene Symbol:
          LOC100129917
          Gene Name:
          uncharacterized LOC100129917
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 780149 - 781848 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00291414_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100129917
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_036511.1 1 48 74
          GenBank mRNA AK055583.1 - 1 50 74
          BC040652.2 - 1 36 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400012885.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC04000954.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK055583.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 61813 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 165518 AF039687.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 165518 AK093783.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 165518 AK314281.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 165518 AX748304.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 165518 BC020794.2 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 165518 BC041351.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 165518 BC064364.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 165518 BC073981.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 165518 BC107764.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 165518 BX640804.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 165518 BX640807.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 165518 BX648598.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 165518 NM_004713.3 refseq_mrna_human_NM
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P110291 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 35737 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 19113 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P110291 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P110291 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00906536 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00906536 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1559528_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0400012885.hg.1 ENST00000503185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012885.hg.1 ENST00000503185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012885.hg.1 ENST00000503185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012885.hg.1 ENST00000503185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012885.hg.1 ENST00000503185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012885.hg.1 ENST00000503185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012885.hg.1 ENST00000503185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012885.hg.1 ENST00000503185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012885.hg.1 ENST00000503405.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012885.hg.1 ENST00000503405.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012885.hg.1 ENST00000503405.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012885.hg.1 ENST00000503405.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012885.hg.1 ENST00000507446.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012885.hg.1 ENST00000507446.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012885.hg.1 NR_036511.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012885.hg.1 NR_036511.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012885.hg.1 NR_036511.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012885.hg.1 NR_036511.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012885.hg.1 NR_036511.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012885.hg.1 NR_036511.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012885.hg.1 NR_036512.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012885.hg.1 NR_036512.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012885.hg.1 NR_036512.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012885.hg.1 NR_036512.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04012829.hg.1 NR_036511.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04012829.hg.1 TC04000954.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04012830.hg.1 ENST00000503185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04012830.hg.1 NR_036511.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04012830.hg.1 NR_036512.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04012830.hg.1 TC04000954.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04012828.hg.1 NR_036511.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04012828.hg.1 TC04000954.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0400189536.hg.1 NR_036511.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400189536.hg.1 TC0400012885.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400189536.hg.1 NR_036511.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012885.hg.1 ENST00000503185.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012885.hg.1 ENST00000503185.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012885.hg.1 ENST00000503185.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012885.hg.1 ENST00000503185.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012885.hg.1 ENST00000503185.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012885.hg.1 ENST00000503185.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012885.hg.1 ENST00000503185.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012885.hg.1 ENST00000503185.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012885.hg.1 ENST00000503405.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012885.hg.1 ENST00000503405.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012885.hg.1 ENST00000503405.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012885.hg.1 ENST00000503405.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012885.hg.1 ENST00000507446.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012885.hg.1 ENST00000507446.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012885.hg.1 NR_036511.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012885.hg.1 NR_036511.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012885.hg.1 NR_036511.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012885.hg.1 NR_036511.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012885.hg.1 NR_036511.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012885.hg.1 NR_036511.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012885.hg.1 NR_036512.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012885.hg.1 NR_036512.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012885.hg.1 NR_036512.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012885.hg.1 NR_036512.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400189535.hg.1 ENST00000503405.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400189535.hg.1 NR_036511.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400189535.hg.1 TC0400012885.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400189535.hg.1 ENST00000503405.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400189535.hg.1 NR_036511.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400189537.hg.1 ENST00000503185.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400189537.hg.1 ENST00000503185.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400189537.hg.1 ENST00000503405.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400189537.hg.1 ENST00000507446.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400189537.hg.1 NR_036511.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400189537.hg.1 NR_036512.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400189537.hg.1 TC0400012885.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400189537.hg.1 ENST00000503185.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400189537.hg.1 ENST00000503185.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400189537.hg.1 ENST00000503405.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400189537.hg.1 ENST00000507446.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400189537.hg.1 NR_036511.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400189537.hg.1 NR_036512.1 Refseq
          Human Genome U133 Plus 2.0 Array 1559528_at - -
          Human Genome U133 Plus 2.0 Array 1559528_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559528_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559528_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559528_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559528_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559528_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559528_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559528_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559528_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559528_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559528_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559528_at6 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A023825 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A023825 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 229050 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3187713 AK055583.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3187713 BC040652.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3187713 NR_036511.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0