Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00294990_m1
          See other LINC01127 GE Assays ›
          Gene Symbol:
          LINC01127
          Gene Name:
          long intergenic non-protein coding RNA 1127
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 101983465 - 101985545 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00294990_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC01127
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK055877.1 - 1 - 2 115 89

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Recommended Assay Homo sapiens Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365150 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365126 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468730 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365173 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365182 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365211 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468663 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365124 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365249 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365187 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365220 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365244 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365143 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468646 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365142 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468691 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468724 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365224 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468726 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468723 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365164 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365163 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365153 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365229 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468616 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365160 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365198 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365178 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468684 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15293114 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365235 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468614 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468618 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365190 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365243 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365174 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468631 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468676 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468752 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365180 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365228 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468756 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365199 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365213 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365186 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15293109 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365135 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365172 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365181 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365192 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365117 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365203 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468697 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365118 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468654 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365157 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468636 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468703 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365188 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365168 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468713 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468704 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365136 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365155 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365227 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365238 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468688 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365226 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365122 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365185 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365212 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365215 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365246 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365167 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365132 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365161 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15293105 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365171 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365204 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365236 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468711 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365137 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365193 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365241 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468638 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468709 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468699 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365234 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365141 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468740 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468645 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365237 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468718 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468725 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365148 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365207 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365230 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15293111 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21468712 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365183 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365231 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365139 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365140 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365144 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01365176 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3479449 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P750164 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 9360 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 22785 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3479449 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P750164 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P750164 - -
          Agilent Human 1B Microarray G4111A A_32_P884716 - -
          Agilent Human 1B Microarray G4111A A_32_P879268 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1560679_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000625702.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000625702.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000625702.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000625702.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000625702.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000625702.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000625702.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627273.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627273.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627273.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627273.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000627337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000628502.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000628502.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000628502.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 ENST00000628502.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 NR_103791.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 NR_103791.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 NR_103791.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 NR_103791.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 NR_103791.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 NR_103791.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008661.hg.1 NR_103791.1 Refseq
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000625702.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000625702.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000625702.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000625702.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000625702.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000625702.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000625702.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627215.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627273.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627273.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627273.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627273.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627337.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627337.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627337.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627337.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000627337.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000628502.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000628502.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000628502.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 ENST00000628502.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 NR_103791.1 Refseq
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 NR_103791.1 Refseq
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 NR_103791.1 Refseq
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 NR_103791.1 Refseq
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 NR_103791.1 Refseq
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 NR_103791.1 Refseq
          Affymetrix Human Clariom D Assay TC0200008661.hg.1 NR_103791.1 Refseq
          Human Genome U133 Plus 2.0 Array 1560679_at - -
          Human Genome U133 Plus 2.0 Array 1560679_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560679_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560679_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560679_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560679_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560679_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560679_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560679_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560679_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560679_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560679_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560679_at4 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0